Skip to Content
Merck
All Photos(2)

Key Documents

203793

Sigma-Aldrich

Mercury(II) oxide

99.999% trace metals basis

Synonym(s):

Mercuric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HgO
CAS Number:
Molecular Weight:
216.59
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: mercury

mp

500 °C (dec.) (lit.)

SMILES string

O=[Hg]

InChI

1S/Hg.O

InChI key

UKWHYYKOEPRTIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junwen Shi et al.
Analytica chimica acta, 583(1), 84-91 (2007-03-28)
In order to investigate trace mercury-containing proteins in maternal rat and their offspring, a method of enriched stable isotopic tracer (196Hg and 198Hg) combined with size-exclusion chromatography (SEC) coupled to inductively coupled plasma-isotope dilution mass spectrometry (ICP-IDMS) was developed. Prior
Ying Li et al.
Environmental science & technology, 40(20), 6444-6448 (2006-11-24)
A novel silica-titania (SiO2-TiO2) nanocomposite has been developed to effectively capture elemental mercury (Hg0) under UV irradiation. Moisture has been reported to have an important impact on this nanocomposite's performance. In this work, the role of moisture on Hg0 removal
H Suginome et al.
Steroids, 55(8), 353-359 (1990-08-01)
3 beta-Hydroxy-5 alpha-androstan-17-one was transformed into 17-oxa-5 alpha-androstan-3 beta-ol in five steps involving conversion of the 17-ketone via the corresponding lactol to its hypoiodite and thence a regioselective beta-scission under irradiation to give ring D seco iodoformate, from which the
Christina Stadlbauer et al.
Analytical and bioanalytical chemistry, 383(3), 500-508 (2005-06-11)
The potential of laser ablation inductively coupled plasma mass spectrometry for the time-resolved analysis of heavy-metal intoxication in human bodies by analysis of hair is demonstrated. As application, we analyzed forensic samples from one individual after Hg intake and from
Investigation of an unexpected addition reaction that occurs when 2,3:4,5-di-O-isopropylidene-D-ribose diethyl dithioacetal is treated with mercuric oxide and mercuric chloride.
T Sambaiah et al.
The Journal of organic chemistry, 66(12), 4405-4408 (2001-06-09)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service