Skip to Content
Merck
All Photos(1)

Key Documents

202673

Sigma-Aldrich

Arsenic(III) oxide

99.995% trace metals basis

Synonym(s):

ATO, Diarsenic trioxide;, Arsenic trioxide, Arsenous acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
As2O3
CAS Number:
Molecular Weight:
197.84
EC Number:
MDL number:
UNSPSC Code:
12161600
eCl@ss:
38080102
PubChem Substance ID:
NACRES:
NA.22

Assay

99.995% trace metals basis

form

powder

reaction suitability

core: arsenic
reagent type: catalyst

SMILES string

O=[As]O[As]=O

InChI

1S/As2O3/c3-1-5-2-4

InChI key

IKWTVSLWAPBBKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Arsenic(III) oxide (ATO) is an inorganic form of arsenic that is used as a therapeutic compound for treating patients with acute promyelocytic leukemia (APL). Many drug-delivery methods are currently being developed using nanotechnology to overcome its poor bioavailability due to rapid clearance from blood.

Application


  • Electrochemical detection applications: A study on La-based perovskites incorporated with functionalized carbon nanofibers highlights the use of arsenic trioxide for the electrochemical detection of roxarsone in water and food samples, providing a method to improve safety and environmental monitoring (John Felix MA et al., 2024).

  • Antiproliferative effects in cancer research: Research demonstrates the inhibitory effects of arsenic trioxide, combined with metformin, on KG1a cell proliferation, suggesting its potential utility in developing antineoplastic strategies (Huang-Li WH et al., 2024).

  • Environmental remediation: A review on manganese oxide-based materials for arsenic removal discusses the application of arsenic trioxide in environmental remediation, focusing on the effectiveness and advancements in arsenic decontamination technologies (Younas M et al., 2024).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Target Organs

Respiratory system,Cardio-vascular system,Gastrointestinal tract

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Recent advances in arsenic trioxide encapsulated nanoparticles as drug delivery agents to solid cancers.
Akhtar A, et al.
Journal of biomedical research, 31(3), 177-177 (2017)
Silica nanovehicles endow arsenic trioxide with an ability to effectively treat cancer cells and solid tumors.
Zhao Z, et al.
Journal of Materials Chemistry, 2(37), 6313-6323 (2014)
A facile route to core?shell nanoparticulate formation of arsenic trioxide for effective solid tumor treatment.
Zhang Z, et al.
Nanoscale, 8(7), 4373-4380 (2016)
Ruoshi Xu et al.
Bone research, 6, 33-33 (2018-11-28)
How osteoblast cells are induced is a central question for understanding skeletal formation. Abnormal osteoblast differentiation leads to a broad range of devastating craniofacial diseases. Here we have investigated intramembranous ossification during cranial bone development in mouse models of skeletal
Cui Li et al.
Toxicology letters, 219(3), 223-230 (2013-04-02)
Arsenic trioxide (As2O3; ATO) is clinically effective in treating acute promyelocytic leukemia (APL); however, it frequently causes cardiotoxic effects. This study was designed to investigate whether ATO could induce apoptosis of cardiac fibroblasts (CFs) that play very important roles in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service