Skip to Content
Merck
All Photos(3)

Key Documents

202452

Sigma-Aldrich

Poly(ethylene glycol)

average MV 8,000, hydroxyl

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23

product name

Poly(ethylene glycol), average Mv ~8,000, powder (crystalline)

form

powder (crystalline)

Quality Level

mol wt

average Mv ~8,000

viscosity

800 cSt(210 °F)(lit.)

mp

60-63 °C (lit.)

density

1.27 g/mL at 25 °C

Ω-end

hydroxyl

α-end

hydroxyl

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • PEG in Targeted Drug Delivery: Investigating the effects of PEGylation on the surface properties of nanoparticles, which plays a crucial role in targeted drug delivery systems (Shi et al., 2021).
  • PEG′s Role in Nanomedicine: Discussing the immunological properties of PEG and exploring its popular role in nanomedicine formulations, with a focus on immunogenic responses (Shi et al., 2022).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilhan Yu et al.
Journal of personalized medicine, 10(1) (2020-03-08)
Developing a high-efficiency manufacturing system for personalized medicine plays an important role in increasing the feasibility of personalized medication. The purpose of this study is to investigate the feasibility of a new extrusion-based fabrication process for personalized drugs with a
Olivia Tort et al.
Journal of lipid research, 59(11), 2108-2115 (2018-09-15)
Cholesterol efflux (CE) capacity has been inversely associated with atherosclerosis and may provide an insight on inflammation occurring in human immunodeficiency virus (HIV) individuals. We address this by studying CE in HIV patients at different stages of HIV disease progression.
M Patel Geeta et al.
Current drug delivery, 6(2), 159-165 (2009-05-20)
Carbamazepine indicated for the control of epilepsy, undergoes extensive hepatic first-pass metabolism after oral administration. A vaginal dosage form of carbamazepine is not commercially available. Conventional suppository having poor retention in the vaginal tract, as they are removed in a
I L Konorova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 7-9 (1991-07-01)
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced
Idalis Villanueva et al.
Acta biomaterialia, 5(8), 2832-2846 (2009-06-11)
The pericellular matrix (PCM) surrounding chondrocytes is thought to play an important role in transmitting biochemical and biomechanical signals to the cells, which regulates many cellular functions including tissue homeostasis. To better understand chondrocytes interactions with their PCM, three-dimensional poly(ethylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service