Skip to Content
Merck
All Photos(1)

Key Documents

200387

Sigma-Aldrich

Polycaprolactone triol

average Mn ~300

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C[CH2O[CO(CH2)5O]nH]3
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

average Mn ~300

Quality Level

transition temp

softening point 10 °C

density

1.07 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Polycaprolactone triol (PCL-triol) may be used to prepare biodegradable elastomer with citric acid for soft tissue engineering applications. A number of studies report the use of PCL-triol in the preparation of biodegradable polyurethane, which finds a number of uses as biomedical implants.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

424.4 °F - closed cup

Flash Point(C)

218 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Evaluation of in situ curable biodegradable polyurethanes containing zwitterion components.
Adhikari R, et al.
Journal of Materials Science. Materials in Medicine, 21(4), 1081-1089 (2010)
(Citric acid?co?polycaprolactone triol) polyester: A biodegradable elastomer for soft tissue engineering.
Thomas L V and Nair P D
Biomatter, 1(1), 81-90 (2011)
Synthetic and tissue-derived models for studying rigidity effects on invadopodia activity.
Methods in Molecular Biology (2013)
Processing and characterization of elastomeric polycaprolactone triol?citrate coatings for biomedical applications.
Shirazi H S, et al.
Progress in Organic Coatings, 77(4), 821-829 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service