198978
3-Methyl-2-oxopentanoic acid sodium salt
≥98%
Synonym(s):
2-Keto-3-methylvaleric acid sodium salt, 3-Methyl-2-oxopentanoic acid sodium salt, Ketoisoleucine sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
Assay
≥98%
form
powder
mp
204-206 °C (lit.)
functional group
ketone
SMILES string
[Na+].CCC(C)C(=O)C([O-])=O
InChI
1S/C6H10O3.Na/c1-3-4(2)5(7)6(8)9;/h4H,3H2,1-2H3,(H,8,9);/q;+1/p-1
InChI key
SMDJDLCNOXJGKC-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
3-Methyl-2-oxopentanoic acid sodium salt may be used in chemical synthesis studies.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Biochimica et biophysica acta, 1637(1), 119-126 (2003-01-16)
Mitochondrial dysfunction occurs in many neurodegenerative diseases. The alpha-ketoglutarate dehydrogenase complex (KGDHC) catalyzes a key and arguably rate-limiting step of the tricarboxylic acid cycle (TCA). A reduction in the activity of the KGDHC occurs in brains and cells of patients
Analysis of (S)- and (R)-3-methyl-2-oxopentanoate enantiomorphs in body fluids.
Methods in enzymology, 324, 33-39 (2000-09-16)
Pancreas, 20(1), 38-46 (2000-01-12)
Insulin-releasing effects of 2-ketobutyric acid (KB), 2-ketoisocaproic acid (KIC), 2-keto-3-methylvaleric acid (KMV), and 3-phenylpyruvic acid (PP) were examined by using clonal beta cells. Whereas KIC, KMV, and PP dose-dependently initiated insulin secretion and potentiated the effects of 4.2-16.7 mM glucose
Journal of bacteriology, 182(11), 3239-3246 (2000-05-16)
Recently the bkd gene cluster from Enterococcus faecalis was sequenced, and it was shown that the gene products constitute a pathway for the catabolism of branched-chain alpha-keto acids. We have now investigated the regulation and physiological role of this pathway.
Annals of the New York Academy of Sciences, 1042, 272-278 (2005-06-21)
The alpha-ketoglutarate dehydrogenase complex (KGDHC) is a mitochondrial enzyme in the TCA cycle. Inhibition of KGDHC activity by alpha-keto-beta-methyl-n-valeric acid (KMV) is associated with neuron death. However, the effect of KMV in microglia is unclear. Therefore, we investigated the effect
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service