Skip to Content
Merck
All Photos(3)

Documents

181110

Sigma-Aldrich

Nylon 6

pellets

Synonym(s):

Polycaprolactam

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-NH(CH2)5CO-]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

pellets

Quality Level

autoignition temp.

815 °F

transition temp

Tm 228.5 °C
Tg 62.5 °C

density

1.084 g/mL at 25 °C (lit.)

InChI

1S/C6H13NO/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H2,7,8)

InChI key

ALBYIUDWACNRRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nylon 6 can be used as starting material to prepare:
  • Silica nanoparticles–nylon 6 composites for extraction of hormones from aqueous samples.
  • Polymeric nanofibers by electrospinning method. These nanofibers can be used to fabricate highly efficient nanofilters.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anna Lindqvist et al.
Perception, 41(11), 1373-1391 (2012-01-01)
Twenty participants scaled similarities in odour quality, odour intensity and pleasantness/ unpleasantness of 10 binary and 5 higher-order mixtures of 5 odorous degradation products from the polymer Polyamide 6.6. The perceived odour qualities of all binary mixtures were represented well
Fei Hang et al.
Nanotechnology, 22(36), 365708-365708 (2011-08-17)
A nanomechanical testing set-up is developed by integrating an atomic force microscope (AFM) for force measurements with a scanning electron microscope (SEM) to provide imaging capabilities. Electrospun nanofibers of polyvinyl alcohol (PVA), nylon-6 and biological mineralized collagen fibrils (MCFs) from
Habib Bagheri et al.
Analytica chimica acta, 716, 34-39 (2012-01-31)
A novel solid phase microextraction (SPME) fiber was fabricated by electrospinning method in which a polymeric solution was converted to nanofibers using high voltages. A thin stainless steel wire was coated by the network of polymeric nanofibers. The polymeric nanofiber
Giriprasath Gururajan et al.
Nanoscale, 3(8), 3300-3308 (2011-06-30)
An atomic force microscopy (AFM) probe is successfully utilized as an electrospinning tip for fabricating Nylon-6 nanofibers. The nanometre-size tip enabled controlled deposition of uniform polymeric nanofibers within a 1 cm diameter area. Nylon-6 nanofibers were continuously electrospun at a
Rongchun Zhang et al.
The journal of physical chemistry. A, 116(3), 979-984 (2011-12-22)
The structure and dynamic behavior of mobile components play a significant role in determining properties of solid materials. Herein, we propose a novel real-time spectrum-editing method to extract signals of mobile components in organic solids on the basis of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service