Skip to Content
Merck
All Photos(1)

Key Documents

172839

Sigma-Aldrich

2-Tridecanone

99%

Synonym(s):

Methyl undecyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COCH3
CAS Number:
Molecular Weight:
198.34
Beilstein:
1757402
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

refractive index

n20/D 1.435 (lit.)

bp

134 °C/10 mmHg (lit.)
263 °C (lit.)

mp

24-27 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CCCCCCCCCCCC(C)=O

InChI

1S/C13H26O/c1-3-4-5-6-7-8-9-10-11-12-13(2)14/h3-12H2,1-2H3

InChI key

CYIFVRUOHKNECG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Tridecanone is a component of the essential oil obtained from the leaves of Zanthoxylum alatum.

Application

2-Tridecanone was used to study the impact of soil amendments in onion bulbs.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjay Guleria et al.
TheScientificWorldJournal, 2013, 790580-790580 (2013-06-20)
The essential oil obtained from the fresh leaves of Zanthoxylum alatum was analysed by gas chromatography/mass spectrometry (GC/MS). Fourteen components were identified, and linalool (30.58%), 2-decanone (20.85%), β -fenchol (9.43%), 2-tridecanone (8.86%), β -phellandrene (5.99%), Sabinene (4.82%), and α -pinene
P J Skiba et al.
Insect biochemistry and molecular biology, 24(8), 847-853 (1994-09-01)
A fatty acid chain elongation process is involved in incorporation of saturated and unsaturated fatty acyl-CoA esters into 2-tridecanone and (Z)-10-heptadecen-2-one by Drosophila buzzatii. The microsomal fraction from mature male ejaculatory bulbs is chain-length specific and requires malonyl-CoA (or acetyl-CoA
Persistence of 2-tridecanone on the leaves of seven vegetables.
G F Antonious
Bulletin of environmental contamination and toxicology, 73(6), 1086-1093 (2005-01-28)
George F Antonious
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 48(4), 302-307 (2013-02-05)
A field study was conducted to investigate the impact of soil amendments on concentrations of two volatile organic compounds, 2-undecanone and 2-tridecanone, in onion bulbs. The soil in five plots was mixed with sewage sludge, five plots were mixed with
Yussef F B Braga et al.
Anais da Academia Brasileira de Ciencias, 79(1), 35-39 (2007-04-03)
The effect of 2-tridecanone vapor on the cowpea weevil (Callosobruchus maculatus) development was determined. Seeds of cowpea were infested with adults and exposed to different doses of 2-tridecanone isolated from Pilocarpus microphyllus Stapf ex Holm, a plant species native from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service