Skip to Content
Merck
All Photos(1)

Key Documents

170518

Sigma-Aldrich

1-Naphthyl isocyanate

98%

Synonym(s):

α-Naphthyl isocyanate, 1-Isocyanatonaphthalene, 1-Naphthalene isocyanate, Naphthalen-1-ylisocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NCO
CAS Number:
Molecular Weight:
169.18
Beilstein:
742779
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.6344 (lit.)

bp

267 °C/761 mmHg (lit.)

mp

4 °C (lit.)

solubility

alcohol: soluble
chloroform: soluble
diethyl ether: soluble

density

1.177 g/mL at 25 °C (lit.)

functional group

isocyanate

SMILES string

O=C=Nc1cccc2ccccc12

InChI

1S/C11H7NO/c13-8-12-11-7-3-5-9-4-1-2-6-10(9)11/h1-7H

InChI key

BDQNKCYCTYYMAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1-Naphthyl isocyanate was used in the preparation of cationic aromatic urethane.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Ullrich et al.
Biomedical chromatography : BMC, 15(3), 212-216 (2001-06-08)
A method for determining the enantiomers of 10 therapeutically relevant aminoalcohols using HPLC and precolumn derivatization was developed. Naphthyl isocyanate reacted with racemic aminoalcohols to form urea derivatives which were separated isocratically on a cellulose tris(3,5-dimethylphenylcarbamate) coated silica gel column
Q Yang et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 24(4), 285-289 (1989-01-01)
alpha-Naphthyl isocyanate (NI) as a derivatizing agent to separate several beta-blockers was described. This procedure is simple, rapid and gives good resolution. Experiments with NI derivatives of alprenolol showed that the isocyanate group reacted with the amino group on the
Christopher J McEntyre et al.
Analytica chimica acta, 644(1-2), 90-94 (2009-05-26)
Choline is an essential nutrient which is difficult to measure because it has no native absorbance or fluorescence and only relatively unreactive functional groups. The method described here uses the reaction of the hydroxyl group on choline with 1-naphthyl isocyanate
Shashi K Ramaiah et al.
Toxicologic pathology, 35(6), 757-766 (2007-10-19)
Polymorphonuclear leukocytes (neutrophils) are essential in the defense against invading microorganisms, tissue trauma or any inciting inflammatory signals. Hepatic infiltration of neutrophils is an acute response to recent or ongoing liver injury, hepatic stress or unknown systemic inflammatory signals. Once
Ying Zhao et al.
Toxicology, 230(2-3), 145-150 (2006-12-23)
A pharmacokinetic study was carried out in rats to investigate the effects of experimental hepatic injury on the liver glucuronidation and bile excretion of ketoprofen (KP) and its glucuronides (KPGs). In vivo, KP (20mg/kg b.w.) was intravenously administered to carbon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service