Skip to Content
Merck
All Photos(2)

Documents

169099

Sigma-Aldrich

Divinylbenzene

technical grade, 55%

Synonym(s):

DVB HP, Diethenylbenzene, Vinylstyrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH=CH2)2
CAS Number:
Molecular Weight:
130.19
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

vapor density

4.5 (vs air)

vapor pressure

0.9 mmHg ( 30 °C)

form

liquid

autoignition temp.

878 °F

contains

≤1500 ppm p-tert-butylcatechol as inhibitor

expl. lim.

0.7 %, 75 °F
6.5 %, 130 °F

concentration

55%

refractive index

n20/D 1.561 (lit.)

bp

195 °C (lit.)

density

0.912 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C=Cc1ccc(C=C)cc1

InChI

1S/C10H10/c1-3-9-7-5-6-8-10(9)4-2/h3-8H,1-2H2

InChI key

MYRTYDVEIRVNKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Divinylbenzene (DVB) has a benzene ring in combination with meta and para isomers of DVB and ethylvinylbenzene. It acts as a monomeric unit which facilitates the preparation of polydivinylbenzene by anionic polymerization.

Application

DVB can be used as a crosslinking agent in the synthesis of silicon/carbon composite for use as an anode material for lithium ion batteries. It can also be used as a monomer to synthesize dimer particles by seed polymerization method for potential uses in electrical clutches and shock absorbers.

Other Notes

contains 4-ethylvinylbenzene and 3-ethylvinylbenzene

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis of silicon/carbon, multi-core/shell microspheres using solution polymerization for a high performance Li ion battery.
Shin K, et al.
Electrochimica Acta, 58(4), 578-582 (2011)
Improved rate capability of lithium-ion batteries with Ag nanoparticles deposited onto silicon/carbon composite microspheres as an anode material.
Kwon E, et al.
Solid State Ionics, 237(4), 28-33 (2013)
Synthesis and Characterization of Linear Poly (divinylbenzene-co-ethylvinylbenzene) via a Cationic Solid Catalyst.
Bensaada N, et al.
Orient. J. Chem., 29(4), 1631-1635 (2014)
Monodisperse conducting colloidal dipoles with symmetric dimer structure for enhancing electrorheology properties.
Shin K, et al.
Journal of Colloid and Interface Science, 374(1), 18-24 (2012)
Sensitive detection of 2, 4, 6-trinitrotoluene by tridimensional monitoring of molecularly imprinted polymer with optical fiber and five-branched gold nanostars.
Cennamo N, et al.
Sensors and Actuators B, Chemical, 208(4), 291-298 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service