Skip to Content
Merck
All Photos(2)

Key Documents

167525

Sigma-Aldrich

Phenyl chloroformate

99%

Synonym(s):

Chloroformic acid phenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOOC6H5
CAS Number:
Molecular Weight:
156.57
Beilstein:
606778
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1 (vs air)

Quality Level

vapor pressure

1.22 psi ( 20 °C)
1.67 psi ( 55 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.511 (lit.)

bp

74-75 °C/13 mmHg (lit.)

density

1.248 g/mL at 25 °C (lit.)

functional group

chloro
phenoxy

storage temp.

2-8°C

SMILES string

ClC(=O)Oc1ccccc1

InChI

1S/C7H5ClO2/c8-7(9)10-6-4-2-1-3-5-6/h1-5H

InChI key

AHWALFGBDFAJAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetics of spontaneous hydrolysis of phenyl chloroformate has been studied in water-ethylene glycol, cationic, zwitterionic, nonionic and anionic micellar solutions.

Application

Phenyl chloroformate was used in the synthesis of poly(2-(phenoxycarbonyloxy)ethyl methacrylate) and phenyl-(4-vinylphenyl) carbonate.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional polymethacrylates as bacteriostatic polymers.
Adelmann R, et al.
Eur. Polymer J., 45(11), 3093-3107 (2009)
Amalia Rodríguez et al.
Langmuir : the ACS journal of surfaces and colloids, 20(23), 9945-9952 (2004-11-03)
The spontaneous hydrolysis of phenyl chloroformate was studied in water-ethylene glycol, EG, cationic, zwitterionic, nonionic, and anionic micellar solutions, the surfactants being tetradecyltrimethylammonium bromide, tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, tricosaoxyethylene glycol ether, and sodium dodecyl sulfate. The dependence of the observed rate constant on
Synthesis and ligand recognition of paracetamol selective polymers: semi-covalent versus non-covalent molecular imprinting.
Rosengren-Holmberg JP, et al.
Organic & Biomolecular Chemistry, 7(15), 3148-3155 (2009)
R K Keller et al.
Biochemistry, 20(20), 5831-5836 (1981-09-29)
A sensitive assay is described for quantitating dolichyl phosphate (Dol-P), the polyprenyl phospholipid which participates in N-linked glycosylation. The assay is based on a novel reaction of alkyl phosphates with phenyl chloroformate in which a monosubstituted mixed anhydride of phosphoric
Luis García-Río et al.
Langmuir : the ACS journal of surfaces and colloids, 21(17), 7672-7679 (2005-08-11)
A study was carried out on the solvolysis of substituted phenyl chloroformates in AOT/isooctane/water microemulsions. (AOT is the sodium salt of bis(2-ethyhexyl)sulfosuccinate.) The results obtained have been interpreted by taking into account the distribution of the chloroformates between the continuous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service