Skip to Content
Merck
All Photos(1)

Key Documents

162329

Sigma-Aldrich

3-Fluorotoluene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4F
CAS Number:
Molecular Weight:
110.13
Beilstein:
1903631
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.469 (lit.)

bp

115 °C (lit.)

mp

−87 °C (lit.)

density

0.991 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Cc1cccc(F)c1

InChI

1S/C7H7F/c1-6-3-2-4-7(8)5-6/h2-5H,1H3

InChI key

BTQZKHUEUDPRST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reaction of 3-fluorotoluene with excesses of methanol has been investigated on the large-pore catalysts and on the medium-pore HZSM-5.

Application

3-Fluorotoluene was used in gas phase selective synthesis of 3-fluorobenzaldehyde. It was used in generation of tolyl cations by chemical ionization in mass spectrometry.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

David M Marcus et al.
Langmuir : the ACS journal of surfaces and colloids, 20(14), 5946-5951 (2006-02-08)
The reactions of fluorobenzene, 3-fluorotoluene, and three isomers of difluorotoluene, chlorobenzene, and bromobenzene with excesses of methanol were investigated on the large-pore catalysts HBeta (*BEA) and HSAPO-5 (AFI), and on the medium-pore HZSM-5 (MFI). Flow reactor studies in pulse mode
Vapor phase oxidation of 4-fluorotoluene over vanadia-titania catalyst.
Maurya SK, et al.
J. Mol. Catal. A: Chem., 234(1), 51-57 (2005)
T G Heath et al.
Journal of the American Society for Mass Spectrometry, 2(4), 270-277 (1991-08-01)
The ion/molecule reaction of the tolyl cation with dimethyl ether has been investigated using triple quadrupole mass spectrometry. Three isomers with [C7H7](+) composition, the 3-tolyl, benzyl, and tropylium cations, were individually selected and reacted with dimethyl ether at a pressure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service