Skip to Content
Merck
All Photos(1)

Key Documents

159166

Sigma-Aldrich

1,3-Benzodioxole

99%

Synonym(s):

1,2-(Methylenedioxy)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12
Beilstein:
115506
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

12 mmHg ( 25 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.539 (lit.)

bp

172-173 °C (lit.)

density

1.064 g/mL at 25 °C (lit.)

SMILES string

C1Oc2ccccc2O1

InChI

1S/C7H6O2/c1-2-4-7-6(3-1)8-5-9-7/h1-4H,5H2

InChI key

FTNJQNQLEGKTGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Supersonic jet fluorescence spectra of 1,3-benzodioxole has been studied. The far-infrared spectrum of the vapour of 1,3-benzodioxole has been reported. The electronic absorption spectra and the laser-induced fluorescence spectra of supersonic-jet-cooled 1,3-benzodioxole molecules has been investigated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie-Josée Haglund Halsør et al.
Scientific reports, 10(1), 11995-11995 (2020-07-21)
Nonulosonic acid (NulO) biosynthesis in bacteria is directed by nab gene clusters that can lead to neuraminic, legionaminic or pseudaminic acids. Analysis of the gene content from a set mainly composed of Aliivibrio salmonicida and Moritella viscosa strains reveals the
1, 3-benzodioxole: far-infrared spectrum 50-500 cm< sup>- 1</sup>.
Duckett JA, et al.
Chemical Physics Letters, 64(2), 261-265 (1979)
Supersonic jet fluorescence spectroscopy of 1, 3-benzodioxole: A non-planar (C< sub> 2</sub>) structure in S0.
Hassan KM and Hollas JM.
Chemical Physics Letters, 157(3), 183-188 (1989)
Spectroscopic Determination of the Vibrational Potential Energy Surface and Conformation of 1, 3-Benzodioxole in Its S1 (p, p*) Excited State. The Effect of the Electronic Excitation on the Anomeric Effect.
Laane J, et al.
Journal of the American Chemical Society, 122(11), 2628-2634 (2000)
Ron Amon et al.
Oncotarget, 8(68), 112236-112244 (2018-01-20)
Humans have circulating antibodies against diverse glycans containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service