Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
158968
α-D-Glucose
anhydrous, 96%
Synonym(s):
alpha-D-Glucose, alpha-Glucose, α-Dextrose
About This Item
Recommended Products
grade
anhydrous
Quality Level
Assay
96%
form
powder or crystals
optical activity
[α]20/D +52°, c = 10 in H2O
mp
153-156 °C (lit.)
SMILES string
OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1
InChI key
WQZGKKKJIJFFOK-DVKNGEFBSA-N
Gene Information
human ... PYGM(5837)
Looking for similar products? Visit Product Comparison Guide
Application
- As a reducing agent in the preparation superparamagnetic ferrous oxide (Fe3O4) nanoparticles[1] and silver nanocrystals.[2]
- As an additive for the formation of isoporous polystyrene-block-poly(4-vinylpyridine) (PS-b-P4VP) diblock copolymer membranes.[3]
- For glycosylation of cell-penetrating poly(disulfide)s (CPDs) with improved solubility to achieve multifunctional cellular uptake.[4]
- As a precursor in the synthesis of metal/carbon nanohybrids under hydrothermal conditions.[5]
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
How to sterilize the product for cell culture after resuspend it?
1 answer-
This product may be filter sterilized using a 0.22 um filter membrane. It may also be autoclaved, however this may cause slight discoloration. Please see the link below to review the product datasheet for Plant Cell Culture tested item G7021, an identical product of another brand. The information may be applied to product 158968.
https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/111/451/g7021pis.pdfHelpful?
-
-
貴社のアルファ・グルコース製品をOGTTとして50-75g投与して、MRスペクトロスコピーで計測しようと考えています。ヒトに投与しても安全でしょうか?
1 answer-
Unless otherwise stated this product is intended for research use only and is not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses, or any type of consumption or application to humans or animals.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service