Skip to Content
Merck
All Photos(3)

Key Documents

158402

Sigma-Aldrich

Dicyclohexano-18-crown-6

98%

Synonym(s):

2,3,11,12-Dicyclohexano-1,4,7,10,13,16-hexaoxacyclooctadecane, 2,5,8,15,18,21-Hexaoxatricyclo[20.4.0.09.14]hexacosane, Dicyclohexyl-18-crown-6, Perhydro-dibenzo-18-crown-6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H36O6
CAS Number:
Molecular Weight:
372.50
Beilstein:
1130529
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

functional group

ether

SMILES string

C1CCC2OCCOCCOC3CCCCC3OCCOCCOC2C1

InChI

1S/C20H36O6/c1-2-6-18-17(5-1)23-13-9-21-11-15-25-19-7-3-4-8-20(19)26-16-12-22-10-14-24-18/h17-20H,1-16H2

InChI key

BBGKDYHZQOSNMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Anion activator, and complexing agent which solubilizes alkali metal ions in non-polar solvents.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Toshiyuki Fujii et al.
The journal of physical chemistry. A, 114(7), 2543-2552 (2010-02-04)
This work reports on the chemical isotope fractionation of Zn(II) by a solvent extraction method with the crown ether dicyclohexano-18-crown-6. The (m)Zn/(64)Zn ratios (m = 66, 67, and 68) were analyzed by multiple-collector inductively coupled plasma mass spectrometry. The relative
Mahmoud Chamsaz et al.
Journal of hazardous materials, 167(1-3), 597-601 (2009-02-24)
A simple single drop liquid-phase microextraction (SDME) technique, combined with electrothermal atomic absorption spectroscopy (ETAAS) is developed both to preconcentrate and determine thallium (I) ions in aqueous solutions. The ions were transferred from 10.0 ml of aqueous sample (donor phase)
Hitoshi Mizuguchi et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 27(1), 85-89 (2011-01-15)
A simple and inexpensive preconcentration technique has been developed for the ultra-trace determination of lead(II) using electrothermal atomic absorption spectrometry (ETAAS). The lead(II) complex with dicyclohexano-18-crown 6-ether (DC18C6) was extracted to a small piece of cellulose acetate-type membrane filter (2
Radioiodination of alkyl halides with Na 125I catalyzed by dicyclohexyl-18-crown-6.
L Bo-Li et al.
Radioisotopes, 30(7), 391-393 (1981-07-01)
Negar Bahraini et al.
Health physics, 101(2), 128-135 (2011-06-29)
A molecularly imprinted polymer (MIP) comprising dicyclohexano-18-crown-6 (DCH18C6) was synthesized as a Sr-selective sorbent for urine bioassay purposes. MIP particles (326 ± 2 nm diameter) were formed using acetone and acetonitrile (1:3 v/v) as the porogen, methacrylic acid (MAA) as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service