Skip to Content
Merck
All Photos(1)

Key Documents

157430

Sigma-Aldrich

2-Ethylbutyryl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2CHC(O)Cl
CAS Number:
Molecular Weight:
134.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

137-139 °C (lit.)

density

0.982 g/mL at 25 °C (lit.)

SMILES string

CCC(CC)C(Cl)=O

InChI

1S/C6H11ClO/c1-3-5(4-2)6(7)8/h5H,3-4H2,1-2H3

InChI key

SMUKODJVMQOSAB-UHFFFAOYSA-N

Application

2-Ethylbutyryl chloride was used in syntheses of bicyclic thiophene derivatives.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph L Duffy et al.
Bioorganic & medicinal chemistry letters, 15(5), 1401-1405 (2005-02-17)
A novel class of antagonists of the human glucagon receptor (hGCGR) has been discovered. Systematic modification of the lead compound identified substituents that were essential for activity and those that were amenable to further optimization. This SAR exploration resulted in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service