15580
Boron
≥95% (boron), amorphous powder
Synonym(s):
B
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
B
CAS Number:
Molecular Weight:
10.81
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Assay
≥95% (boron)
form
amorphous powder
reaction suitability
core: boron
reagent type: catalyst
resistivity
1.5E12 μΩ-cm, 20 °C
particle size
≤1 μm
density
2.34 g/mL at 25 °C (lit.)
SMILES string
[B]
InChI
1S/B
InChI key
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Boron (amorphous powder) has been used as a boron source to synthesize hexagonal boron nitride (h-BN), boron doped diamond (BDD) and europium-doped BN nanotubes.
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Aluminium oxynitride?hexagonal boron nitride composites with anisotropic properties.
Wilk A, et al.
J. Eur. Ceram. Soc., 36(8), 2087-2092 (2016)
Boron doped diamond synthesized from detonation nanodiamond in a COH fluid at high pressure and high temperature.
Shakhov F M, et al.
Journal of Solid State Chemistry, 256, 72-92 (2017)
Eu?doped Boron Nitride Nanotubes as a Nanometer?Sized Visible?Light Source.
Chen H, et al.
Advanced Materials, 19(14), 1845-1848 (2017)
Yu Zhao et al.
Journal of the American Chemical Society, 135(4), 1201-1204 (2013-01-16)
Two kinds of boron and nitrogen co-doped carbon nanotubes (CNTs) dominated by bonded or separated B and N are intentionally prepared, which present distinct oxygen reduction reaction (ORR) performances. The experimental and theoretical results indicate that the bonded case cannot
Tara A Devirian et al.
Critical reviews in food science and nutrition, 43(2), 219-231 (2003-04-23)
Boron may be an essential nutrient for animals and humans. Dietary boron influences the activity of many metabolic enzymes, as well as the metabolism of steroid hormones and several micronutrients, including calcium, magnesium, and vitamin D. Boron supplementation in rats
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service