151637
Isoxazole
99%
Synonym(s):
1,2-Oxazole, 1-Oxa-2-azacyclopentadiene, 2-Azafuran
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H3NO
CAS Number:
Molecular Weight:
69.06
Beilstein:
103773
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
2.4 (vs air)
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.427 (lit.)
bp
93-95 °C (lit.)
density
1.078 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
c1cnoc1
InChI
1S/C3H3NO/c1-2-4-5-3-1/h1-3H
InChI key
CTAPFRYPJLPFDF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Isoxazole are described as inhibitors of acetylcholinesterase (AChE). Isoxazole ligands bind to and inhibit the Sxc- antiporter.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
53.6 °F - closed cup
Flash Point(C)
12 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Adam A Wallace et al.
The journal of physical chemistry. A, 124(38), 7768-7775 (2020-08-28)
We report a photoelectron imaging study of gas-phase deprotonation of isoxazole in which spectroscopic data are compared to the results of electronic structure calculations for the anion products corresponding to each of three possible deprotonation sites. The observed photoelectron spectra
Afnan A Matti et al.
Bioorganic & medicinal chemistry letters, 23(21), 5931-5935 (2013-09-18)
Microwave accelerated reaction system (MARS) technology provided a good method to obtain selective and open isoxazole ligands that bind to and inhibit the Sxc- antiporter. The MARS provided numerous advantages, including: shorter time, better yield and higher purity of the
Nikolai V Rostovskii et al.
The Journal of organic chemistry, 82(1), 256-268 (2016-12-16)
4-Aminopyrrole-3-carboxylates and pyrazine-2-carboxylates were synthesized from 5-alkoxyisoxazoles and 1-sulfonyl-1,2,3-triazoles by tuning the Rh(II) catalyst and the reaction conditions. The reaction in chloroform at 100 °C under Rh
Margarita Gutiérrez et al.
The Journal of pharmacy and pharmacology, 65(12), 1796-1804 (2013-11-05)
Inhibition of acetylcholinesterase (AChE) is a common treatment for early stages of Alzheimer's disease. In this study, nine isoxazoles derivatives were tested for their in-vitro AChE activity. The molecular docking showed the interaction of the compounds with the active site.
Adam A Wallace et al.
The journal of physical chemistry. A, 125(1), 317-326 (2020-12-29)
Electron capture by the σ* LUMO of isoxazole triggers the dissociation of the O-N bond and the opening of the ring. Photodetachment of the resulting anion accesses a neutral structure, in which the O· and ·N bond fragments interact through
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service