Skip to Content
Merck
All Photos(2)

Key Documents

151076

Sigma-Aldrich

9-Borabicyclo[3.3.1]nonane solution

0.5 M in THF

Synonym(s):

9-BBN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15B
CAS Number:
Molecular Weight:
122.02
Beilstein:
605509
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

0.5 M in THF

density

0.894 g/mL at 25 °C

SMILES string

B1C2CCCC1CCC2

InChI

1S/C8H15B/c1-3-7-5-2-6-8(4-1)9-7/h7-9H,1-6H2/t7-,8+

InChI key

FEJUGLKDZJDVFY-OCAPTIKFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

9-Borabicyclo[3.3.1]nonane is an organic compound, which is generally used as a hydroborating agent. It is thermally stable, less sensitive to oxygen and water when compared to other dialkyl boranes. It is commonly used to prepare aldehydes from alkynes.

Application

Differentiation of symmetrical diols by reduction of the corresponding cyclic acetals with 9-BBN.
Protecting group for alkenes†

Reactant for:
  • Linear SPPS synthesis of ubiquitin derivatives
  • Copper-catalyzed cross-coupling reactions of organoboron compounds with primary alkyl halides and pseudohalides
  • Intramolecular insertion of alkenes into palladium-nitrogen bonds
  • Preparation of (phosphonoacetyl)ornithine to study effect on arginine biosynthetic genes in yeast
  • Hetero-Diels-Alder reaction for synthesis of spirocyclic alkaloids
Selective hydroboration reduction reagent.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and properties
Hydroboration and Organic Synthesis: 9-Borabicyclo [3.3.1] nonane (9-BBN) (2007)
Zhishan Bo et al.
Organic letters, 6(5), 667-669 (2004-02-28)
Several useful aryl/alkyl building blocks for dendrimer and hyperbranched polymer synthesis were prepared by Suzuki-Miyaura cross-coupling reactions and iododesilylation reactions. The iododesilylations were carried out with iodine monochloride (ICl) in methylene chloride and diethyl ether (10:1). This solvent combination lowered
9-Borabicyclo [3.3. 1] nonane as a convenient selective hydroborating agent
Knights EF and Brown HC
Journal of the American Chemical Society, 90(19), 5281-5283 (1968)
Chemistry Letters (Jpn), 33, 396-397 (2004)
Chemtracts, 13, 219-222 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service