Skip to Content
Merck
All Photos(3)

Key Documents

143936

Sigma-Aldrich

Propionamide

97%

Synonym(s):

Propanamide, Propylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CONH2
CAS Number:
Molecular Weight:
73.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

bp

213 °C (lit.)

mp

76-79 °C (lit.)

solubility

alcohol: freely soluble
chloroform: freely soluble
diethyl ether: freely soluble
water: freely soluble

density

1.042 g/mL at 25 °C (lit.)

functional group

amide

SMILES string

CCC(N)=O

InChI

1S/C3H7NO/c1-2-3(4)5/h2H2,1H3,(H2,4,5)

InChI key

QLNJFJADRCOGBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propionamide on γ-irradiation reacts with sulfur dioxide and this reaction has been studied by ESR spectroscopy, gas absorption measurements and X-ray diffraction[1].

Application

Propionamide was used as adsorbent in the determination of adsorption isotherms of acetamide and propionamide on multi-wall carbon nanotube[2]. It was used in a robust screening method to study biotransformations using (+)-γ-lactamase enzyme[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of Adsorption Isotherms of Acetamide and Propionamide on Carbon Nanotube.
Vadi M and Moradi N.
Orient. J. Chem., 27(4), 1491-1491 (2011)
Reactions of Gases with Irradiated Organic Solids: III. Reactions of Propionamide, n-Butyramide, Isobutyramide, Methacrylamide, Valeramide, and Stearamide with Sulfur Dioxide.
Perotti A, et al.
Mol. Cryst. Liq. Cryst., 9(1), 323-342 (2969)
Dale W Sickles et al.
Toxicology and applied pharmacology, 222(1), 111-121 (2007-06-02)
The microtubule (MT) motor protein kinesin is a vital component of cells and organs expressing acrylamide (ACR) toxicity. As a mechanism of its potential carcinogenicity, we determined whether kinesins involved in cell division are inhibited by ACR similar to neuronal
M R Wilkins et al.
Journal of molecular biology, 289(3), 645-657 (1999-06-05)
The availability of genome sequences, affordable mass spectrometers and high-resolution two-dimensional gels has made possible the identification of hundreds of proteins from many organisms by peptide mass fingerprinting. However, little attention has been paid to how information generated by these
Richard H Stadler
Advances in experimental medicine and biology, 561, 157-169 (2006-01-28)
This paper summarizes the progress made to date on acrylamide research pertaining to analytical methods, mechanisms of formation, and mitigation research in the major food categories. Initial difficulties with the establishment of reliable analytical methods have today in most cases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service