Skip to Content
Merck
All Photos(3)

Documents

140112

Sigma-Aldrich

4-Nitrobenzyl chloride

99%

Synonym(s):

α-Chloro-4-nitrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2Cl
CAS Number:
Molecular Weight:
171.58
Beilstein:
387187
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

70-73 °C (lit.)

solubility

chloroform: soluble 50 mg/mL, clear to very slightly hazy, faintly yellow

SMILES string

[O-][N+](=O)c1ccc(CCl)cc1

InChI

1S/C7H6ClNO2/c8-5-6-1-3-7(4-2-6)9(10)11/h1-4H,5H2

InChI key

KGCNHWXDPDPSBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitrobenzyl chloride acts as substrate for glutathione S-transferase(GST) in determination of GST in Chinese fetal liver. It undergoes reduction by NADPH to yield 4-nitrotoluene.

Application

4-Nitrobenzyl chloride was used to prepare unsymmetrically N,N′-bis(substituted) 4,13-diaza-18-crown-6-ether derivatives.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Jemth et al.
The Journal of biological chemistry, 275(12), 8618-8624 (2000-03-18)
Ser(11) in rat glutathione transferase T2-2 is important for stabilization of the reactive enzyme-bound glutathione thiolate in the reaction with 1-menaphthyl sulfate. The S11A mutation increased the pK(a) value for the pH dependence of the rate constant for pre-steady-state product
Tetrahedron Letters, 34, 95-95 (1993)
S B Lei et al.
Zhongguo yao li xue bao = Acta pharmacologica Sinica, 11(5), 389-391 (1990-09-01)
Subcellular fractions were isolated from Chinese fetal liver at 4-8 months of age for the determination of glutathione S-transferase (GST). Using 1-chloro-2,4-dinitrobenzene (CDNB) as substrate, GST activity was found to be 66 +/- 34 nmol/(min.mg protein), mainly in the cytosol.
S N Moreno et al.
The Journal of biological chemistry, 261(17), 7811-7815 (1986-06-15)
The o-, m-, and p-nitrobenzyl chlorides are reduced aerobically and anaerobically by NADPH and rat hepatic microsomes. Under aerobic conditions, these nitro anion radicals reduce oxygen to superoxide as demonstrated by oxygen consumption and spin trapping of superoxide with 5,5-dimethyl-1-pyrroline
Shingo Arakawa et al.
Drug metabolism and disposition: the biological fate of chemicals, 40(3), 497-503 (2011-12-16)
We investigated the impact of glutathione transferases Mu 1 (GSTM1)- and glutathione transferase Theta 1 (GSTT1)-null genotypes on hepatic GST activities in humans and compared the results with those of Gstm1- and Gstt1-null mice. In liver with GSTM1/Gstm1-null genotype, GST

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service