All Photos(1)
About This Item
Linear Formula:
CH3(CH2)6CN
CAS Number:
Molecular Weight:
125.21
Beilstein:
1744063
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.42 (lit.)
bp
198-200 °C (lit.)
mp
−45 °C (lit.)
density
0.814 g/mL at 25 °C (lit.)
functional group
nitrile
SMILES string
CCCCCCCC#N
InChI
1S/C8H15N/c1-2-3-4-5-6-7-8-9/h2-7H2,1H3
InChI key
YSIMAPNUZAVQER-UHFFFAOYSA-N
Application
Heptyl cyanide (Caprylonitrile) was used as a solvent to study the intermediates in the decomposition of aliphatic diazo-compounds.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
165.2 °F - closed cup
Flash Point(C)
74 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Intermediates in the decomposition of aliphatic diazo-compounds. Part XI. Mechanistic studies on the reaction of diphenylmethylene with amines in solution.
Bethell D, et al.
J. Chem. Soc. Perkin Trans. II, 11, 1307-1312 (1974)
Peter S Toth et al.
The Analyst, 140(6), 1947-1954 (2015-02-11)
Two-phase voltammetry has been carried out using a reverse cell configuration, i.e. with the lower density organic solvent on the top of the aqueous solution in the cell, where the organic solvents contain either nitrile or ketone functional groups. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service