Skip to Content
Merck
All Photos(2)

Key Documents

126217

Sigma-Aldrich

2,4-Dihydroxybenzophenone

99%

Synonym(s):

DHB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3COC6H5
CAS Number:
Molecular Weight:
214.22
Beilstein:
1311566
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

powder

mp

144.5-147 °C (lit.)

SMILES string

Oc1ccc(c(O)c1)C(=O)c2ccccc2

InChI

1S/C13H10O3/c14-10-6-7-11(12(15)8-10)13(16)9-4-2-1-3-5-9/h1-8,14-15H

InChI key

ZXDDPOHVAMWLBH-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Application


  • Substituent Effects on the Ultraviolet Absorption Properties of 2, 4-Dihydroxy Dibenzophenone: This research examines how different substituents affect the UV absorption characteristics of 2,4-dihydroxybenzophenone, a compound important for its UV protective properties (Wu et al., 2022).

  • Kevlar® and Nomex modification via 2, 4-dihydroxybenzophenone anchoring improves water repellency and induces antibacterial and UV protection properties: Explores the enhancement of Kevlar and Nomex fabrics by anchoring 2,4-dihydroxybenzophenone to improve their functional properties (Tonis et al., 2023).

Legal Information

Kevlar is a registered trademark of E. I. du Pont de Nemours and Company

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

257.0 °F

Flash Point(C)

125 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalie C Bamford et al.
Nature communications, 11(1), 2450-2450 (2020-05-18)
The exopolysaccharide galactosaminogalactan (GAG) is an important virulence factor of the fungal pathogen Aspergillus fumigatus. Deletion of a gene encoding a putative deacetylase, Agd3, leads to defects in GAG deacetylation, biofilm formation, and virulence. Here, we show that Agd3 deacetylates
Kresten Ole Kusk et al.
Environmental toxicology and chemistry, 30(4), 959-966 (2011-01-05)
Benzophenone (BP)-type ultraviolet (UV) filters are widely used in cosmetic and sunscreen products and can enter the aquatic environment. Therefore, we investigated the subchronic toxicity of 2,4-dihydroxybenzophenone (BP1) on the marine calanoid copepod Acartia tonsa in an early life-stage development
Ting-Hao Kuo et al.
Analytica chimica acta, 1080, 95-103 (2019-08-15)
Structural analysis of biomolecules is essential to natural product discovery, especially for precious biomaterials such as agarwood. However, one of the greatest challenges to the characterization of natural products is the profound cost in time and manpower to the structural
Sarah J Hanna et al.
Physical chemistry chemical physics : PCCP, 11(36), 7963-7975 (2009-09-04)
An investigation of oleic acid and 2,4-dihydroxybenzoic (DHB) acid aerosols was carried out using an aerosol mass spectrometer with pulsed lasers for vaporization and ionization and an ion trap for mass analysis. The extent of ion fragmentation was studied as
Fatma Zohra Djazouli Alim et al.
PloS one, 14(6), e0216679-e0216679 (2019-06-19)
The "ship" of the Arabian and North African deserts, the one-humped dromedary camel (Camelus dromedarius) has a remarkable capacity to survive in conditions of extreme heat without needing to drink water. One of the ways that this is achieved is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service