12072
Benzamidine
≥95.0%
Synonym(s):
Benzenylamidine, Phenylamidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5C(=NH)NH2
CAS Number:
Molecular Weight:
120.15
Beilstein:
606020
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95.0%
form
powder or crystals
functional group
amine
phenyl
SMILES string
NC(=N)c1ccccc1
InChI
1S/C7H8N2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H3,8,9)
InChI key
PXXJHWLDUBFPOL-UHFFFAOYSA-N
Gene Information
human ... F10(2159) , F2(2147) , PLAT(5327) , PLAU(5328) , PLG(5340) , PRSS1(5644)
Related Categories
Application
Benzamidine was used in the preparation of heparan sulfate-rich proteoglycan from mouse mammary epithelial cells. It has been used to prevent enzymatic degradation during the purification procedure of bovine factor XII (Hageman factor).
Biochem/physiol Actions
Benzamidine is an inhibitor of plasmin and has been used in the radioimmunoassay of glucagon in human plasma. It prevents the destruction of Thyrotropin-releasing hormone (TRH) added to whole blood in vitro before radioimmunoassay of TRH.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Purification and characterization of bovine coagulation factor XII (Hageman factor).
H Claeys et al.
European journal of biochemistry, 87(1), 69-74 (1978-06-01)
Hung-Wei Chang et al.
PLoS pathogens, 15(6), e1007826-e1007826 (2019-06-21)
Vaccinia mature virus requires A26 envelope protein to mediate acid-dependent endocytosis into HeLa cells in which we hypothesized that A26 protein functions as an acid-sensitive membrane fusion suppressor. Here, we provide evidence showing that N-terminal domain (aa1-75) of A26 protein
Fatma Rmili et al.
Biotechnology progress, 35(4), e2833-e2833 (2019-05-03)
A mesophilic bacterial culture, producing an extracellular alkaline lipase, was isolated from the gas-washing wastewaters generated from the Sfax phosphate plant of the Tunisian Chemical Group and identified as Staphylococcus capitis strain. The lipase, named S. capitis lipase (SCL), has
Use of benzamidine as a proteolytic inhibitor in the radioimmunoassay of glucagon in plasma.
J W Ensinck et al.
The Journal of clinical endocrinology and metabolism, 35(3), 463-467 (1972-09-01)
Ana L Roa-Espitia et al.
Biology open, 5(9), 1189-1199 (2016-07-13)
Several focal adhesion proteins are known to cooperate with integrins to link the extracellular matrix to the actin cytoskeleton; as a result, many intracellular signaling pathways are activated and several focal adhesion complexes are formed. However, how these proteins function
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service