Skip to Content
Merck
All Photos(1)

Key Documents

115568

Sigma-Aldrich

(S)-(−)-α-Methylbenzylamine

98%

Synonym(s):

(S)-(-)-alpha-Methylbenzylamine, (S)-(−)-1-Phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
2204907
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.5 mmHg ( 20 °C)

Quality Level

Assay

98%

optical activity

[α]20/D −39°, neat

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.526 (lit.)

bp

187 °C (lit.)

density

0.94 g/mL at 25 °C (lit.)

functional group

amine
phenyl

storage temp.

2-8°C

SMILES string

C[C@H](N)c1ccccc1

InChI

1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3/t7-/m0/s1

InChI key

RQEUFEKYXDPUSK-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(-)-α-Methylbenzylamine along with 2-formylphenylboronic acid has been used in a derivatization protocol to analyze the enantiomeric excess of chiral diols.
It can also be used:
  • In the diastereoselective synthesis of S-aminonitriles.
  • As a chiral auxiliary in the synthesis of (S)-(-)-N-acetylcalycotomine or (R)-(+)-N-acetylcalycotomine.
  • As a chiral building block in the asymmetric synthesis of 1-substituted tetrahydro-β-carbolines.
Used in a one-pot, multi-component synthesis of a highly substituted, chiral pyrrole.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashwin R Bharadwaj et al.
Organic letters, 6(14), 2465-2468 (2004-07-02)
[reaction: see text] A multicomponent synthesis of highly substituted pyrroles catalyzed by thiazolium salts has been disclosed. The reaction employs an acyl anion conjugate addition reaction of acylsilanes (sila-Stetter) and unsaturated ketones to generate 1,4-dicarbonyl compounds in situ. The subsequent
(S)-(-)-a-Methylbenzylamine as an efficient chiral auxiliary in enantiodivergent synthesis of both enantiomers of N-acetylcalycotomine.
Ziolkowski M, et al
Tetrahedron Asymmetry, 10(17), 3371-3380 (1999)
Lithium perchlorate/diethylether catalyzed aminocyanation of aldehydes
Heydari A, et al
Tetrahedron Letters, 39(19), 3049-3050 (1998)
Paul E Harrington et al.
Current medicinal chemistry, 14(28), 3027-3034 (2008-01-29)
The calcium sensing receptor (CaR) is a G protein-coupled receptor (GPCR) that plays a fundamental role in serum calcium homeostasis. The CaR is expressed on the chief cells of the parathyroid gland and is responsible for controlling the secretion of
Bartosz Lewandowski et al.
Chemical communications (Cambridge, England), (47)(47), 6399-6401 (2008-12-03)
Simple chiral aza-crown ethers based on sucrose display high enantioselectivity in complexation of phenylethylammonium chlorides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service