Skip to Content
Merck
All Photos(1)

Key Documents

111775

Sigma-Aldrich

3-Methyl-2-pentene, mixture of cis and trans

98%

Synonym(s):

1,2,3-Trimethylpropene, 2-Ethyl-2-butene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)=CHCH3
CAS Number:
Molecular Weight:
84.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.404 (lit.)

bp

69 °C (lit.)

density

0.698 g/mL at 25 °C (lit.)

SMILES string

CC\C(C)=C\C

InChI

1S/C6H12/c1-4-6(3)5-2/h4H,5H2,1-3H3/b6-4+

InChI key

BEQGRRJLJLVQAQ-GQCTYLIASA-N

Application

3-Methyl-2-pentene (mixture of cis and trans) is used in the study of photosensitized oxidation of trialkylalkenes in the internal framework of Na-ZSM-5 zeolites.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Zhe Chen et al.
The Journal of organic chemistry, 70(12), 4676-4681 (2005-06-04)
Photosensitized oxidation of trialkylalkenes 2-methyl-2-pentene (1), 1-methylcyclohexene (2), trans-3-methyl-2-pentene (3), cis-3-methyl-2-pentene (4), and 2-methyl-2-butene (5) included in the internal framework of Na-ZSM-5 zeolites was investigated. The zeolite samples having adsorbed the alkenes were suspended in isooctane, and the sensitizer, tetraphenylporphyrin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service