Skip to Content
Merck
All Photos(1)

Key Documents

110388

Sigma-Aldrich

Butylcyclohexane

≥99%

Synonym(s):

1-Cyclohexylbutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3C6H11
CAS Number:
Molecular Weight:
140.27
Beilstein:
1900631
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

2.9 mmHg ( 37.7 °C)

Quality Level

Assay

≥99%

autoignition temp.

475 °F

refractive index

n20/D 1.441 (lit.)

bp

178-180 °C (lit.)

mp

−78 °C (lit.)

density

0.818 g/mL at 25 °C (lit.)

SMILES string

CCCCC1CCCCC1

InChI

1S/C10H20/c1-2-3-7-10-8-5-4-6-9-10/h10H,2-9H2,1H3

InChI key

GGBJHURWWWLEQH-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

Butylcyclohexane reacts with n-butylbenzene by free-radical mechanism to form 1-methylcyclohexene and cyclohexane. Butylcyclohexane, when photo-oxidized by solar light on synthetic seawater, forms cyclohexanone.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photosensitized oxidation of n-butylcyclohexane as a model for photochemical degradation of n-alkylcyclohexanes in seawater.
Journal of Photochemistry and Photobiology A: Chemistry, 46(3), 347-355 (1989)
Thermal Decomposition of Jet Fuel Model Compounds under Near-Critical and Supercritical Conditions. 1. n-Butylbenzene and n-Butylcyclohexane.
Industrial & Engineering Chemistry Research, 37(12), 4591-4600 (1998)
Carmen M Dominguez et al.
Journal of environmental management, 261, 110240-110240 (2020-03-10)
Chlorinated pesticides were extensively produced in the XX century, generating high amounts of toxic wastes often dumped in the surroundings of the production sites, resulting in hot points of soil and groundwater pollution worldwide. This is the case of Bailín
Jae-Man Park et al.
Nature communications, 11(1), 4638-4638 (2020-09-17)
Existing gels are mostly polar, whose nature limits their role in soft devices. The intermolecular interactions of nonpolar polymer-liquid system are typically weak, which makes the gel brittle. Here we report highly soft and transparent nonpolar organogels. Even though their
Emre Seyyal et al.
Analytica chimica acta, 964, 96-111 (2017-03-30)
Principles of sol-gel chemistry were utilized to create silica- and germania-based dual-ligand surface-bonded sol-gel coatings providing enhanced performance in capillary microextraction (CME) through a combination of ligand superhydrophobicity and π-π interaction. These organic-inorganic hybrid coatings were prepared using sol-gel precursors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service