Skip to Content
Merck
All Photos(1)

Documents

10914

Sigma-Aldrich

Isopropenyl acetate

produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)

Synonym(s):

IPA, 1-Methylvinyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C(CH3)=CH2
CAS Number:
Molecular Weight:
100.12
Beilstein:
1280347
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

produced by Wacker Chemie AG, Burghausen, Germany

Assay

≥99.0% (GC)

refractive index

n20/D 1.401 (lit.)

bp

94 °C (lit.)

density

0.922 g/mL at 20 °C
0.909 g/mL at 25 °C (lit.)

SMILES string

CC(=C)OC(C)=O

InChI

1S/C5H8O2/c1-4(2)7-5(3)6/h1H2,2-3H3

InChI key

HETCEOQFVDFGSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopropenyl acetate is a superior acetylating agent and decomposes into ketene and acetone by heating in presence of acid catalysts. Iodine in isopropenyl acetate is a unique catalyst for the acetylation of a variety of alcohols, phenols and amines under solvent free conditions.

Other Notes

prices for bulk quantities on request

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of isopropenyl acetate.
Hagemeyer HJ and Hull DC.
Ind. and Eng. Chem., 41(12), 2920-2924 (1949)
Molecular iodine in isopropenyl acetate (IPA): a highly efficient catalyst for the acetylation of alcohols, amines and phenols under solvent free conditions.
Ahmed N and van Lier JE.
Tetrahedron Letters, 47(30), 5345-5349 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service