Skip to Content
Merck
All Photos(1)

Key Documents

10906

Sigma-Aldrich

Chloroacetaldehyde dimethyl acetal

produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)

Synonym(s):

CADMA, 2-Chloro-1,1-dimethoxyethane, Dimethyl chloroacetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH(OCH3)2
CAS Number:
Molecular Weight:
124.57
Beilstein:
1733700
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

produced by Wacker Chemie AG, Burghausen, Germany

Quality Level

Assay

≥99.0% (GC)

refractive index

n20/D 1.415 (lit.)

bp

128-130 °C (lit.)

density

1.094 g/mL at 25 °C (lit.)

SMILES string

COC(CCl)OC

InChI

1S/C4H9ClO2/c1-6-4(3-5)7-2/h4H,3H2,1-2H3

InChI key

CRZJPEIBPQWDGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroacetaldehyde dimethyl acetal is a clear, colorless liquid. It reacts with hydroxythiol to form hydroxy acetal.

Application

Chloroacetaldehyde dimethyl acetal can be used as a reactant to synthesize:
  • 2-(chloromethyl)-4,7-dethyl-1,3-dioxepane
  • 2-(chloromethyl)-5,6-benzo-1,3-dioxepane
  • ketene dimethyl acetal

Chloroacetaldehyde dimethyl acetal was used to investigate the enzymes used for activation and transformation of vinyl chloride. It was also used for introducing O-(2,2-dimethoxyethyl) groups into amylose, dextran, and a linear (1→3)-β-D-glucan. Chloroacetaldehyde dimethyl acetal was utilised in the synthesis of ethoxyacetylide.

Other Notes

prices for bulk quantities on request

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coupling of polysaccharides activated by means of chloroacetaldehyde dimethyl acetal to amines or proteins by reductive amination.
B?gwald J, et al.
Carbohydrate Research, 148(1), 101-107 (1986)
Recent trends in the synthesis of carbazoles: an update.
Tetrahedron, 68(31), 6099-6121 (2012)
Camphor derived 1, 4-oxathianes for carbonyl epoxidation.
Tetrahedron Asymmetry, 9(10), 1801-1807 (1998)
Activation of vinyl chloride to covalently bound metabolites: roles of 2-chloroethylene oxide and 2-chloroacetaldehyde.
F P Guengerich et al.
Biochemistry, 18(23), 5177-5182 (1979-11-13)
Simple enols. 3. Stereochemistry of simple enols in solution.
Capon B
The Journal of Organic Chemistry, 49(2), 255-257 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service