Skip to Content
Merck
All Photos(1)

Documents

108510

Sigma-Aldrich

4-Bromobenzoic acid

98%

Synonym(s):

p-Bromobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CO2H
CAS Number:
Molecular Weight:
201.02
Beilstein:
1906923
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

252-254 °C (lit.)

solubility

ethanol: soluble 5%
H2O: slightly soluble (hot)
alcohol: slightly soluble
diethyl ether: slightly soluble

SMILES string

OC(=O)c1ccc(Br)cc1

InChI

1S/C7H5BrO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)

InChI key

TUXYZHVUPGXXQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Berit Packert Jensen et al.
Rapid communications in mass spectrometry : RCM, 19(4), 519-524 (2005-01-25)
Inductively coupled plasma mass spectrometry (ICPMS) has been used to determine the rate and routes of excretion of bromine following the intraperitoneal administration (50 mg kg(-1)) of 2-, 3- and 4-bromobenzoic acids to male bile-duct-cannulated rats. Analysis of urine and
Quantification of the in vitro and in vivo metabolic fates of 2-, 3-and 4-bromobenzoic acids using high temperature LC coupled to ICP-MS and linear ion trap MS.
Smith CJ, et al.
Chromatographia, 67(9-10), 673-678 (2008)
Ved Vati Singh et al.
Dalton transactions (Cambridge, England : 2003), 44(2), 725-732 (2014-11-20)
The metastable tetragonal Cu2Se phase as nanoflakes has been synthesized for the first time by treating CuCl2 taken in a mixture (1:1) of 1-octadecene and oleylamine with H2N-(CH2)3-SePh dissolved in 1-octadecene. Powder X-ray diffraction (PXRD), HRTEM, SEM-EDX and XPS have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service