10783
Antimony(III) sulfate
≥95.0%
Synonym(s):
Antimony trisulfate, Diantimony trisulfate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Sb2(SO4)3
CAS Number:
Molecular Weight:
531.71
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder
Recommended Products
grade
for analytical purposes
Assay
≥95.0%
form
powder
reaction suitability
reagent type: catalyst
core: antimony
SMILES string
[Sb+3].[Sb+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
InChI
1S/3H2O4S.2Sb/c3*1-5(2,3)4;;/h3*(H2,1,2,3,4);;/q;;;2*+3/p-6
InChI key
MVMLTMBYNXHXFI-UHFFFAOYSA-H
Related Categories
Application
Antimony(III) sulfate may be used as a catalyst for the imino Diels-Alder reaction of Schiff′s bases with N-vinylpyrrolidin-2-one to yield 2-aryl-4-(20 -oxopyrrolidinyl-10 )-1,2,3,4- tetrahydroquinolines.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Imino Diels-Alder Reactions: Efficient Synthesis of 2-Aryl-4-(29-oxopyrrolidinyl-19)-1,2,3,4-tetrahydroquinolines catalysed by Antimony(III) Sulfate.
Srinivasa A, et al.
Monatshefte fur Chemie / Chemical Monthly, 139(3), 255-259 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service