Skip to Content
Merck
All Photos(1)

Documents

104906

Sigma-Aldrich

Dimethoxydimethylsilane

95%

Synonym(s):

DMDMS, Dimethyldimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si(OCH3)2(CH3)2
CAS Number:
Molecular Weight:
120.22
Beilstein:
1733694
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

impurities

3% methyl alcohol

refractive index

n20/D 1.369 (lit.)

bp

81.4 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

CO[Si](C)(C)OC

InChI

1S/C4H12O2Si/c1-5-7(3,4)6-2/h1-4H3

InChI key

JJQZDUKDJDQPMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dimethoxydimethylsilane (DMDMS) can be used:
  • To synthesize various macroporous silica aerogels.
  • In the preparation of hollow silica nanoparticles loaded with phthalocyanine for near-infrared photodynamic and photothermal combination therapy.
  • To synthesize dielectric silicone elastomer with high dielectric permittivity.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new soft dielectric silicone elastomer matrix with high mechanical integrity and low losses.
Madsen F B, et al.
Royal Society of Chemistry Advances, 5(14), 10254-10259 (2015)
Facile synthesis of marshmallow?like macroporous gels usable under harsh conditions for the separation of oil and water.
Hayase G, et al.
Angewandte Chemie (International Edition in English), 52(7), 1986-1989 (2013)
Hydrophobic and physical properties of the ambient pressure dried silica aerogels with sodium silicate precursor using various surface modification agents.
Rao A P, et al.
Applied Surface Science, 253(14), 6032-6040 (2007)
Hila Monder et al.
Polymers, 12(10) (2020-10-25)
The wettability of poly (dimethylsiloxane) (PDMS) coating on plasma-treated glass was studied at room temperature using polar and non-polar liquids. The wettability was investigated regarding the liquids' surface tensions (STs), dielectric constants (DCs) and solubility parameters (SPs). For polar liquids
Juanjuan Peng et al.
Biomaterials, 34(32), 7905-7912 (2013-07-31)
Owing to the convenience and minimal invasiveness, phototherapy, including photodynamic therapy (PDT) and photothermal therapy (PTT), is emerging as a powerful technique for cancer treatment. To date, however, few examples of combination PDT and PTT have been reported. Phthalocyanine (Pc)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service