Skip to Content
Merck
All Photos(1)

Documents

104582

Sigma-Aldrich

2-Bromo-2′,5′-dimethoxyacetophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3COCH2Br
CAS Number:
Molecular Weight:
259.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

83-85 °C (lit.)

SMILES string

COc1ccc(OC)c(c1)C(=O)CBr

InChI

1S/C10H11BrO3/c1-13-7-3-4-10(14-2)8(5-7)9(12)6-11/h3-5H,6H2,1-2H3

InChI key

RGQNFYVSEWDUEI-UHFFFAOYSA-N

General description

2-Bromo-2′,5′-dimethoxyacetophenone can be used in composing phase change inks.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service