103322
N-Methylcyclohexylamine
99%
Synonym(s):
N-Cyclohexylmethylamine
About This Item
Recommended Products
vapor density
4 (vs air)
vapor pressure
11 mmHg ( 40 °C)
Assay
99%
form
liquid
refractive index
n20/D 1.456 (lit.)
bp
149 °C (lit.)
mp
−9-−7 °C (lit.)
density
0.868 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CNC1CCCCC1
InChI
1S/C7H15N/c1-8-7-5-3-2-4-6-7/h7-8H,2-6H2,1H3
InChI key
XTUVJUMINZSXGF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
95.0 °F
Flash Point(C)
35 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service