Merck
  • Zyxin regulates endothelial von Willebrand factor secretion by reorganizing actin filaments around exocytic granules.
Zyxin regulates endothelial von Willebrand factor secretion by reorganizing actin filaments around exocytic granules.
Nature communications (2017-03-04)
Xiaofan Han, Pin Li, Zhenghao Yang, Xiaoshuai Huang, Guoqin Wei, Yujie Sun, Xuya Kang, Xueting Hu, Qiuping Deng, Liangyi Chen, Aibin He, Yingqing Huo, Dong Li, Eric Betzig, Jincai Luo
ABSTRACT
Endothelial exocytosis of Weibel-Palade body (WPB) is one of the first lines of defence against vascular injury. However, the mechanisms that control WPB exocytosis in the final stages (including the docking, priming and fusion of granules) are poorly understood. Here we show that the focal adhesion protein zyxin is crucial in this process. Zyxin downregulation inhibits the secretion of von Willebrand factor (VWF), the most abundant cargo in WPBs, from human primary endothelial cells (ECs) induced by cAMP agonists. Zyxin-deficient mice exhibit impaired epinephrine-stimulated VWF release, prolonged bleeding time and thrombosis, largely due to defective endothelial secretion of VWF. Using live-cell super-resolution microscopy, we visualize previously unappreciated reorganization of pre-existing actin filaments around WPBs before fusion, dependent on zyxin and an interaction with the actin crosslinker α-actinin. Our findings identify zyxin as a physiological regulator of endothelial exocytosis through reorganizing local actin network in the final stage of exocytosis.
Product Description
MISSION® shRNA Product Offerings, Order Custom and Predesigned shRNA
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture