Merck
  • A-kinase anchoring protein BIG3 coordinates oestrogen signalling in breast cancer cells.
A-kinase anchoring protein BIG3 coordinates oestrogen signalling in breast cancer cells.
Nature communications (2017-05-31)
Tetsuro Yoshimaru, Masaya Ono, Yoshimi Bando, Yi-An Chen, Kenji Mizuguchi, Hiroshi Shima, Masato Komatsu, Issei Imoto, Keisuke Izumi, Junko Honda, Yasuo Miyoshi, Mitsunori Sasa, Toyomasa Katagiri
ABSTRACT
Approximately 70% of breast cancer cells express oestrogen receptor alpha (ERα). Previous studies have shown that the Brefeldin A-inhibited guanine nucleotide-exchange protein 3-prohibitin 2 (BIG3-PHB2) complex has a crucial role in these cells. However, it remains unclear how BIG3 regulates the suppressive activity of PHB2. Here we demonstrate that BIG3 functions as an A-kinase anchoring protein that binds protein kinase A (PKA) and the α isoform of the catalytic subunit of protein phosphatase 1 (PP1Cα), thereby dephosphorylating and inactivating PHB2. E2-induced PKA-mediated phosphorylation of BIG3-S305 and -S1208 serves to enhance PP1Cα activity, resulting in E2/ERα signalling activation via PHB2 inactivation due to PHB2-S39 dephosphorylation. Furthermore, an analysis of independent cohorts of ERα-positive breast cancers patients reveal that both BIG3 overexpression and PHB2-S39 dephosphorylation are strongly associated with poor prognosis. This is the first demonstration of the mechanism of E2/ERα signalling activation via the BIG3-PKA-PP1Cα tri-complex in breast cancer cells.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-LMNB1 antibody produced in mouse, clone 4B10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Ser/Thr Phosphatase Assay Kit 1 (K-R-pT-I-R-R), Ser/Thr Phosphatase Assay Kit 1used to detect PP2A activity by either dephosphorylation of the phosphopeptide.