Merck
  • Comparison of phase structures and surface free energy values for the coatings synthesised from linear polyurethanes and from waterborne polyurethane cationomers.
Comparison of phase structures and surface free energy values for the coatings synthesised from linear polyurethanes and from waterborne polyurethane cationomers.
Colloid and polymer science (2011-12-02)
Piotr Król, Bożena Król, Kinga Pielichowska, Stanisław Pikus
ABSTRACT
WAXS, DSC and AFM methods were employed to compare phase structures of the coatings obtained from waterborne polyurethane cationomers which had been synthesised in the reaction of some diisocyanates (MDI, IPDI, TDI and HDI) with polyoxyethylene glycols (M = 600 and 2,000) and butane1,4-diol or N-methyl- or N-butyldiethanolamine and 2,2,3,3-tetrafluoro-1,4-butanediol. The structures were also analysed of the coatings derived from linear polyurethanes which had been synthesised on the basis of similar raw materials. Better rigidity was found for generally amorphous cationomer coats. Changes were discussed in the surface free energy (SFE) values and in their components, as calculated independently with the use of the van Oss-Good and Owens-Wendt methods. Polyurethane coats turned out more hydrophobic as compared to cationomer ones. In both coat types, fluorine incorporated into cationomers contributed to lower SFE values: from 50 down to about 30 mJ/m(2).
Sigma-Aldrich
Isophorone diisocyanate, 98%, mixture of isomers
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N-Phenyldiethanolamine, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2,2,3,3-Tetrafluoro-1,4-butanediol