Merck
Synthesis and Structure of the Zintl Phase SrSn(3)Sb(4).
Inorganic chemistry (1997-08-13)
Donna T. Chow, Robert McDonald, Arthur Mar
ABSTRACT
The new Zintl compound strontium tin antimonide, SrSn(3)Sb(4), has been synthesized, and its structure has been determined by single-crystal X-ray diffraction methods. It crystallizes in the orthorhombic space group -Pnma with a = 10.060(2) Å, b = 4.361(1) Å, c = 22.934(4) Å, and Z = 4 at 22 degrees C. The structure of SrSn(3)Sb(4) may be described as consisting of large, narrow channels running along [010] defined by 30-membered rings constructed from an anionic framework of linked SnSb(4) tetrahedra, SnSb(3) trigonal pyramids, and zigzag Sb-Sb chains. Sr(2+) cations are situated within these channels. The channel structures of SrSn(3)Sb(4) and Ba(2)Sn(3)Sb(6) are closely related.
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Antimony, beads, ≤5 mm, low oxide, 99.999% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Strontium, beads, ≤2.0 mm particle size, 99.99% trace metals basis