Skip to Content
Merck
All Photos(1)

Key Documents

441899

Sigma-Aldrich

Strontium

dendritic pieces, purified by distillation, 99.99% trace metals basis

Synonym(s):

Strontium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sr
CAS Number:
Molecular Weight:
87.62
EC Number:
MDL number:
UNSPSC Code:
12141504
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.99% trace metals basis

form

dendritic pieces

purified by

distillation

reaction suitability

reagent type: reductant

resistivity

23 μΩ-cm, 20°C

bp

1384 °C (lit.)

mp

757 °C (lit.)

density

2.6 g/mL at 25 °C (lit.)

SMILES string

[Sr]

InChI

1S/Sr

InChI key

CIOAGBVUUVVLOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Strontium is a divalent alkaline earth metal with a silvery luster. It is an active reducing agent and readily reacts with oxygen, sulfur, and halogen. It is widely used in the preparation of Sr-based optical materials for piezoelectric, electro-optic devices, and bioactive glasses for bone regeneration.

Application

Applied in the solid state synthesis of a Zintl phase, SrSn3Sb4, with an anionic channel framework of 30-membered rings.[1]
As strontium can enhance the replication of preosteoblastic cells and stimulates bone formation, it can be used as a precursor in the preparation of bioactive glasses for bone regeneration.
Used to make Strontium-potassium-niobate (Sr2KNb5O15, SKN). SKN is of great interest for pyroelectric sensors and various electro-optic devices.2

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Irrit. 2 - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zuzana Saidak et al.
Pharmacology & therapeutics, 136(2), 216-226 (2012-07-24)
Osteoporosis is an important age-related bone disease characterized by increased bone turnover with insufficient bone formation relative to bone resorption. According to the current understanding of this disorder, anti-resorptive and anabolic drugs have been developed for therapeutic intervention. Another therapeutic
Emily Y Tsui et al.
Nature chemistry, 5(4), 293-299 (2013-03-21)
Redox-inactive metals are found in biological and heterogeneous water oxidation catalysts, but, at present, their roles in catalysis are not well understood. Here, we report a series of high-oxidation-state tetranuclear-dioxido clusters comprising three manganese centres and a redox-inactive metal (M).
Strontium and bone.
W E Cabrera et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 14(5), 661-668 (1999-05-13)
R G Robinson et al.
JAMA, 274(5), 420-424 (1995-08-02)
To present the current state of systemic radiopharmaceutical therapy for the palliation of pain in individuals with metastatic cancer and to evaluate the palliative effect and degree of hemotoxicity of strontium chloride 89 (89Sr) in patients with painful osteoblastic metastases
Faisal Hammad Mekky Koua et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(10), 3889-3894 (2013-02-22)
Oxygen-evolving complex of photosystem II (PSII) is a tetra-manganese calcium penta-oxygenic cluster (Mn4CaO5) catalyzing light-induced water oxidation through several intermediate states (S-states) by a mechanism that is not fully understood. To elucidate the roles of Ca(2+) in this cluster and

Questions

1–2 of 2 Questions  
  1. Is this product sealed with Ar or He (inert gas) in glass?

    1 answer
    1. This product is packaged in sealed ampules under Argon.

      Helpful?

  2. Question about 4N Strontium (5g):441899-5G and Calcium (5g):441872-5G. Are they sealed with Ar or He (inert gas) in glass?

    1 answer
    1. Yes, both products are packaged in sealed ampules under Argon.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service