Merck
  • Correlating wine quality indicators to chemical and sensory measurements.
Correlating wine quality indicators to chemical and sensory measurements.
Molecules (Basel, Switzerland) (2015-05-20)
Helene Hopfer, Jenny Nelson, Susan E Ebeler, Hildegarde Heymann
ABSTRACT
Twenty-seven commercial Californian Cabernet Sauvignon wines of different quality categories were analyzed with sensory and chemical methods. Correlations between five quality proxies-points awarded during a wine competition, wine expert scores, retail price, vintage, and wine region-were correlated to sensory attributes, volatile compounds, and elemental composition. Wine quality is a multi-faceted construct, incorporating many different layers. Depending on the quality proxy studied, significant correlations between quality and attributes, volatiles and elements were found, some of them previously reported in the literature.
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
2-Undecanone, 99%
Sigma-Aldrich
Ethyl 2-methylbutyrate, 99%
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl 2-methylbutyrate, ≥98%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ethyl 2-methylbutyrate, natural, mixture of enantiomers, ≥98%, FCC, FG
Sigma-Aldrich
(±)-2-Methylbutyric acid, natural, ≥98%, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
2-Undecanone, natural, FCC, FG
Sigma-Aldrich
2-Undecanone, ≥98%, FCC, FG
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl