Merck
  • Structural study of caveolin-1 intramembrane domain by circular dichroism and nuclear magnetic resonance.
Structural study of caveolin-1 intramembrane domain by circular dichroism and nuclear magnetic resonance.
Biopolymers (2014-12-05)
Guanhua Yang, Zhe Dong, Haoran Xu, Chunyu Wang, Haichao Li, Zhengqiang Li, Fei Li
ABSTRACT
Caveolin-1 is a main structural component of caveolae and essential for the invagination of caveolae by forming a hairpin-shaped structure in the membrane-inserting domain (residues, 102-122). In this article, we determined the tertiary structures of the peptides comprising residues 93-126 and 101-126 of caveolin-1 in 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) aqueous solution and sodium dodecyl sulfate (SDS) micelles, respectively, by nuclear magnetic resonance (NMR) study. The self-association of the peptides in SDS and dodecylphosphocholine (DPC) micelles was also studied by circular dichroism (CD), NMR, and sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) techniques. Our results indicated that both peptides form a helix-break-helix structure with two helices spanning over Leu103-Phe107 and Ile117-His126 and a loop ranging over Gly108-Gly116. The longer peptide 93-126 showed a stronger propensity to aggregate than the shorter peptide 101-126 in the micelles. Our results suggested that the glycine residues at positions 108 and 116 are important for the break of the helical structure of the membrane-inserting domain and the segment Thr93-Arg101 flanking the membrane-inserting domain may play a role in the self-association of the caveolin-1 protein at cellular membrane.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
Sigma-Aldrich
Diphenylcyclopropenone, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Sodium dodecyl-d25 sulfate, ≥98 atom % D, ≥98% (CP)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D