Merck
  • X-ray structurally characterized sensors for ratiometric detection of Zn(2+) and Al(3+) in human breast cancer cells (MCF7): development of a binary logic gate as a molecular switch.
X-ray structurally characterized sensors for ratiometric detection of Zn(2+) and Al(3+) in human breast cancer cells (MCF7): development of a binary logic gate as a molecular switch.
Dalton transactions (Cambridge, England : 2003) (2015-06-09)
Abhijit Ghosh, Debasis Das
ABSTRACT
A very simple molecule derived from salicylaldehyde and N-phenyl ethylenediamine (L1) functions as dual-mode ratiometric fluorescence "turn on" sensors for Zn(2+) and Al(3+) at two different wavelengths. The sensing is based on the combined effect of inhibition of excited-state intra-molecular proton transfer (ESIPT), CH=N isomerization and chelation-enhanced fluorescence (CHEF). Moreover, the [L1-Zn(2+)] system functions as a better Al(3+) sensor where Al(3+) ratiometrically displaces Zn(2+) from the [L1-Zn(2+)] complex. Emission wavelength dependent differentiation of Zn(2+) and Al(3+) using L1 allows us to develop a binary logic gate that functions as a molecular switch. L1 efficiently detects Zn(2+) and Al(3+) in human breast cancer cells (MCF7) while the [L1-Zn(2+)] complex specifically detects Al(3+) in the said cells.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
N-Phenylethylenediamine, 98%
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Salicylaldehyde, reagent grade, 98%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Salicylaldehyde, redist., ≥99.0% (GC)
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS