Skip to Content
Merck
All Photos(3)

Key Documents

S1277

Sigma-Aldrich

Streptomycin sulfate salt

powder, BioXtra, suitable for mouse embryo cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H39N7O12 · 1.5 H2SO4
CAS Number:
Molecular Weight:
728.69
Beilstein:
3894995
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

product line

BioXtra

Quality Level

form

powder

potency

≥720 I.U. per mg

technique(s)

cell culture | embryo: suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.CN[C@H]1[C@H](O)[C@@H](O)[C@H](CO)O[C@H]1O[C@H]2[C@@H](O[C@@H](C)[C@]2(O)C=O)O[C@@H]3[C@@H](O)[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@H]3NC(N)=N.CN[C@H]4[C@H](O)[C@@H](O)[C@H](CO)O[C@H]4O[C@H]5[C@@H](O[C@@H](C)[C@]5(O)C=O)O[C@@H]6[C@@H](O)[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@H]6NC(N)=N

InChI

1S/2C21H39N7O12.3H2O4S/c2*1-5-21(36,4-30)16(40-17-9(26-2)13(34)10(31)6(3-29)38-17)18(37-5)39-15-8(28-20(24)25)11(32)7(27-19(22)23)12(33)14(15)35;3*1-5(2,3)4/h2*4-18,26,29,31-36H,3H2,1-2H3,(H4,22,23,27)(H4,24,25,28);3*(H2,1,2,3,4)/t2*5-,6-,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,21+;;;/m00.../s1

InChI key

QTENRWWVYAAPBI-YZTFXSNBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Streptomycin is used to study mechanisms of streptomycin resistance and is often used together with penicillin and other agents to inhibit bacterial contamination in cell culture applications. It is recommended for use in molecular biology applications at 25-50μg/mL, in cell culture applications at 100 mg/L and in embryo culture at 50 mg/L.

Biochem/physiol Actions

Mode of Action: Streptomycin acts by inhibiting prokaryote protein synthesis by binding to the S12 protein of the 30S ribosomal subunit, thus preventing the transition from imitation complex to chain-elongating ribosome. This causes miscoding or inhibits initiation.

Mode of Resistance: A mutation in rpsL, a gene for S12 ribosomal protein, prevents binding of streptomycin to the ribosome. An aminoglycoside phosphotransferase also inactivates streptomycin.

Antimicrobial spectrum: Streptomycin acts against gram-negative and gram-positive bacteria.

Caution

Solutions can be stored at 2-8°C for up to a month or at -20°C for more extended periods. At 37°C, solutions are stable for three days.

Preparation Note

Stock solutions should be prepared with sterile water at concentrations between 20-25 mg/ml.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Lobenwein et al.
Journal of the American Heart Association, 4(10), e002440-e002440 (2015-10-29)
Paraplegia following spinal cord ischemia represents a devastating complication of both aortic surgery and endovascular aortic repair. Shock wave treatment was shown to induce angiogenesis and regeneration in ischemic tissue by modulation of early inflammatory response via Toll-like receptor (TLR)
Jiayun Zhu et al.
Cell cycle (Georgetown, Tex.), 14(12), 1809-1822 (2015-06-18)
Fanconi Anemia (FA) is a cancer predisposition syndrome and the factors defective in FA are involved in DNA replication, DNA damage repair and tumor suppression. Here, we show that FANCD2 is critical for genome stability maintenance in response to high-linear
Chengfei Zheng et al.
Molecular medicine reports, 12(5), 6656-6662 (2015-09-02)
Aortic aneurysm is a cardiovascular condition with a serious risk of mortality and the dismal prognosis of any type of major cardiovascular disease. The present study found that tissues from aortic aneurysm patients and hypoxic aortic smooth muscle cells showed
Chao Yu et al.
Nature communications, 6, 8017-8017 (2015-08-19)
Oocyte meiosis is a specialized cell cycle that gives rise to fertilizable haploid gametes and is precisely controlled in various dimensions. We recently found that E3 ubiquitin ligase CRL4 is required for female fertility by regulating DNA hydroxymethylation to maintain
Marimuthu Subathra et al.
American journal of physiology. Renal physiology, 309(3), F204-F215 (2015-06-05)
Glycosphingolipids (GSLs) play a role in insulin resistance and diabetes, but their role in diabetic nephropathy (DN) has received limited attention. We used 9- and 17-wk-old nondiabetic db/m and diabetic db/db mice to examine the role of GSLs in DN.

Articles

Antibiotics targeting bacterial ribosomes disrupt protein synthesis, a key process in bacterial growth inhibition.

Antibiotics targeting bacterial ribosomes disrupt protein synthesis, a key process in bacterial growth inhibition.

Antibiotics targeting bacterial ribosomes disrupt protein synthesis, a key process in bacterial growth inhibition.

Antibiotics targeting bacterial ribosomes disrupt protein synthesis, a key process in bacterial growth inhibition.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service