Merck
  • Properdin Contributes to Allergic Airway Inflammation through Local C3a Generation.
Properdin Contributes to Allergic Airway Inflammation through Local C3a Generation.
Journal of immunology (Baltimore, Md. : 1950) (2015-06-28)
Yuan Wang, Takashi Miwa, Blerina Ducka-Kokalari, Imre G Redai, Sayaka Sato, Damodar Gullipalli, James G Zangrilli, Angela Haczku, Wen-Chao Song
ABSTRACT
Complement is implicated in asthma pathogenesis, but its mechanism of action in this disease remains incompletely understood. In this study, we investigated the role of properdin (P), a positive alternative pathway complement regulator, in allergen-induced airway inflammation. Allergen challenge stimulated P release into the airways of asthmatic patients, and P levels positively correlated with proinflammatory cytokines in human bronchoalveolar lavage (BAL). High levels of P were also detected in the BAL of OVA-sensitized and challenged but not naive mice. Compared with wild-type (WT) mice, P-deficient (P(-/-)) mice had markedly reduced total and eosinophil cell counts in BAL and significantly attenuated airway hyperresponsiveness to methacholine. Ab blocking of P at both sensitization and challenge phases or at challenge phase alone, but not at sensitization phase alone, reduced airway inflammation. Conversely, intranasal reconstitution of P to P(-/-) mice at the challenge phase restored airway inflammation to wild-type levels. Notably, C3a levels in the BAL of OVA-challenged P(-/-) mice were significantly lower than in wild-type mice, and intranasal coadministration of an anti-C3a mAb with P to P(-/-) mice prevented restoration of airway inflammation. These results show that P plays a key role in allergen-induced airway inflammation and represents a potential therapeutic target for human asthma.
Sigma-Aldrich
IL-4 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-5 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
IL-5 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-4 from mouse, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture