Skip to Content
Merck

ED

Ethylenediaminetetraacetic acid

purified grade, ≥98.5%, powder

Synonym(s):

(Ethylenedinitrilo)tetraacetic acid, EDTA, Edathamil, Ethylenedinitrilotetraacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(HO2CCH2)2NCH2CH2N(CH2CO2H)2
CAS Number:
Molecular Weight:
292.24
NACRES:
NA.28
PubChem Substance ID:
eCl@ss:
39030907
UNSPSC Code:
12352116
EC Number:
200-449-4
MDL number:
Beilstein/REAXYS Number:
1716295
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purified grade

Quality Level

assay

≥98.5%

form

powder

reaction suitability

reagent type: chelator

pH

2.5 (23 °C, 10 g/L)

mp

250 °C (dec.) (lit.)

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)

InChI key

KCXVZYZYPLLWCC-UHFFFAOYSA-N

General description

Ethylenediaminetetraacetic acid (EDTA) is an affordable chelating agent. It is an aminopolycarboxylic acid and a hexadentate ligand and forms octahedral complex with cations. Ethylenediaminetetraacetic acid (EDTA) is not recommended for phytoremediation as it can leach into natural water resources. It is used in cosmetics and pharmaceutical industry. It triggers platelet aggregation in pseudothrombocytopenia. In chelation therapy, it elicits protection from oxidative damage during blood and lipid peroxidation and may be useful to treat cardiovascular and carbon tetrachloride induced liver fibrosis.

Application

Ethylenediaminetetraacetic acid may be used:
  • as a component of staining buffer
  • as buffer component for incubating epithelial cell adhesion molecule (EpCAM+) cells with fluorescein isothiocyanate (FITC)-conjugated antibody for cell sorting studies
  • for treating epithelial cells for isolation of mucosal myofibroblasts



Still not finding the right product?

Explore all of our products under Ethylenediaminetetraacetic acid


pictograms

Health hazardExclamation mark

signalword

Warning

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves

target_organs

Respiratory Tract

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - STOT RE 2 Inhalation

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Functional modulation of Crohn?s disease myofibroblasts by anti-tumor necrosis factor antibodies
Di Sabatino A, et al.
Gastroenterology, 133(1), 137-149 (2007)
Membrane glucocorticoid receptors (mGCR) are expressed in normal human peripheral blood mononuclear cells and up-regulated after in vitro stimulation and in patients with rheumatoid arthritis
Bartholome B, et al.
Faseb Journal, 18(1), 70-80 (2004)
The phenotypes of pluripotent human hepatic progenitors
Schmelzer E, et al.
Stem Cells, 24(8), 1852-1858 (2006)



Global Trade Item Number

SKUGTIN
M3782-1EA04061824138757
M3907-1EA04061824138788
Z683329-1EA04061833582558
Z683310-1EA04061833582541
SBP2349-0.05MG04061832165783
EDSPAK00104053252468704