Merck
  • An in vitro digestion test that reflects rat intestinal conditions to probe the importance of formulation digestion vs first pass metabolism in Danazol bioavailability from lipid based formulations.
An in vitro digestion test that reflects rat intestinal conditions to probe the importance of formulation digestion vs first pass metabolism in Danazol bioavailability from lipid based formulations.
Molecular pharmaceutics (2014-09-30)
Mette U Anby, Tri-Hung Nguyen, Yan Yan Yeap, Orlagh M Feeney, Hywel D Williams, Hassan Benameur, Colin W Pouton, Christopher J H Porter
ABSTRACT
The impact of gastrointestinal (GI) processing and first pass metabolism on danazol oral bioavailability (BA) was evaluated after administration of self-emulsifying drug delivery systems (SEDDS) in the rat. Danazol absolute BA was determined following oral and intraduodenal (ID) administration of LFCS class IIIA medium chain (MC) formulations at high (SEDDSH-III) and low (SEDDSL-III) drug loading and a lipid free LFCS class IV formulation (SEDDS-IV). Experiments were conducted in the presence and absence of ABT (1-aminobenzotriazole) to evaluate the effect of first pass metabolism. A series of modified in vitro lipolysis tests were developed to better understand the in vivo processing of SEDDS in the rat. Danazol BA was low (<13%) following oral and ID administration of either SEDDS. Increasing drug loading, ID rather than oral administration, and administration of SEDDS-IV rather than SEDDS-III led to higher oral BA. After pretreatment with ABT, however, danazol oral BA significantly increased (e.g., 60% compared to 2% after administration of SEDDSL-III), no effect was observed on increasing drug loading, and differences between SEDDS-III and -IV were minimal. In vitro digestion models based on the lower enzyme activity and lower dilution conditions expected in the rat resulted in significantly reduced danazol precipitation from SEDDS-III or SEDDS-IV on initiation of digestion. At the doses administered here (4-8 mg/kg), the primary limitation to danazol oral BA in the rat was first pass metabolism, and the fraction absorbed was >45% after oral administration of SEDDS-III or SEDDS-IV. In contrast, previous studies in dogs suggest that danazol BA is less dependent on first pass metabolism and more sensitive to changes in formulation processing. In vitro digestion models based on likely rat GI conditions suggest less drug precipitation on formulation digestion when compared to equivalent dog models, consistent with the increases in in vivo exposure (fraction absorbed) seen here in ABT-pretreated rats.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Glyceryl tributyrate, ≥99%
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Ethylene sulfite, 98%
Progesterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glyceryl tributyrate, puriss., ≥98.5% (GC)
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Supelco
Glyceryl tributyrate, analytical standard