Merck
  • Tungsten carbide nanotubes supported platinum nanoparticles as a potential sensing platform for oxalic acid.
Tungsten carbide nanotubes supported platinum nanoparticles as a potential sensing platform for oxalic acid.
Analytical chemistry (2014-07-16)
Thandavarayan Maiyalagan, Palanisamy Kannan, Martin Jönsson-Niedziolka, Joanna Niedziolka-Jönsson
ABSTRACT
Supported tungsten carbide is an efficient and vital nanomaterial for the development of high-performance, sensitive, and selective electrochemical sensors. In this work, tungsten carbide with tube-like nanostructures (WC NTs) supported platinum nanoparticles (PtNPs) are synthesized and explored as an efficient catalyst toward electrochemical oxidation of oxalic acid for the first the time. The WC NTs supported PtNPs modified glassy carbon (GC) electrode is highly sensitive toward the electrochemical oxidation of oxalic acid. A large decrease in the oxidation overpotential (220 mV) and significant enhancement in the peak current compared to unmodified and Pt/C modified GC electrodes have been observed without using any redox mediator. Moreover, WC NTs supported PtNPs modified electrode possessed wide linear concentration ranges from 0 to 125 nM and a higher sensitivity toward the oxidation of oxalic acid (80 nA/nM) achieved by the amperometry method. The present modified electrode showed an experimentally determined lowest detection limit (LOD) of 12 nM (S/N = 3). Further, WC NTs supported PtNPs electrode can be demonstrated to have an excellent selectivity toward the detection of oxalic acid in the presence of a 200-fold excess of major important interferents. The practical application of WC NTs supported PtNPs has also been demonstrated in the detection of oxalic acid in tomato fruit sample, by differential pulse voltammetry under optimized conditions.
Product Description
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, suitable for HPLC
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
Water, suitable for ion chromatography
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I