Merck
  • Semiconductor/relaxor 0-3 type composites without thermal depolarization in Bi₀.₅Na₀.₅TiO₃-based lead-free piezoceramics.
Semiconductor/relaxor 0-3 type composites without thermal depolarization in Bi₀.₅Na₀.₅TiO₃-based lead-free piezoceramics.
Nature communications (2015-03-20)
Ji Zhang, Zhao Pan, Fei-Fei Guo, Wen-Chao Liu, Huanpo Ning, Y B Chen, Ming-Hui Lu, Bin Yang, Jun Chen, Shan-Tao Zhang, Xianran Xing, Jürgen Rödel, Wenwu Cao, Yan-Feng Chen
ABSTRACT
Commercial lead-based piezoelectric materials raised worldwide environmental concerns in the past decade. Bi₀.₅Na₀.₅TiO₃-based solid solution is among the most promising lead-free piezoelectric candidates; however, depolarization of these solid solutions is a longstanding obstacle for their practical applications. Here we use a strategy to defer the thermal depolarization, even render depolarization-free Bi₀.₅Na₀.₅TiO₃-based 0-3-type composites. This is achieved by introducing semiconducting ZnO particles into the relaxor ferroelectric 0.94Bi₀.₅Na₀.₅TiO₃-0.06BaTiO₃ matrix. The depolarization temperature increases with increasing ZnO concentration until depolarization disappears at 30 mol% ZnO. The semiconducting nature of ZnO provides charges to partially compensate the ferroelectric depolarization field. These results not only pave the way for applications of Bi₀.₅Na₀.₅TiO₃-based piezoceramics, but also have great impact on the understanding of the mechanism of depolarization so as to provide a new design to optimize the performance of lead-free piezoelectrics.
Product Description
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate ChemBeads
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC