Skip to Content
Merck
All Photos(5)

Key Documents

S2127

Sigma-Aldrich

Sodium carbonate

Sodium carbonate
1 of 1 reviewers received a sample product or took part in a promotion

ReagentPlus®, ≥99.5%

Synonym(s):

Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5%
form:
powder (granular)
Pricing and availability is not currently available.

Agency

suitable for EPA 300
suitable for SM 2310
suitable for SM 2320

Quality Level

product line

ReagentPlus®

Assay

≥99.5%

form

powder (granular)

pH

12 (25 °C, 106 g/L)

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

mp

851 °C (lit.)

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium carbonate (Na2CO3) is an inorganic compound also known as soda ash. It is used in flue gas desulfurization, and in detergent formulations. It is a stronger alkali used as a softening agent in laundry detergents.

Application

Sodium carbonate can be used:
  • A sintering aid with the ZnO (zinc oxide) for the preparation of BaCe0.5Zr0.3Y0.2O3−δ based composite electrolyte material by solid-state reaction.
  • A co-precipitating agent for the synthesis of stable iron oxide nanoparticles from a stoichiometric combination of Fe(II) and Fe(III) salts.
  • A catalyst for the etherification of glycerol to polyglycerols.

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Palomero-Gallagher et al.
The Journal of comparative neurology, 508(6), 906-926 (2008-04-12)
Anterior cingulate cortex (ACC) is involved in emotion, mood, and autonomic regulation. Although a subgenual part of ACC (sACC) may be vulnerable in depression and area 25 is cytologically unique, there are no assessments that contrast this region with pregenual
Johanna Chiche et al.
Cell metabolism, 29(6), 1243-1257 (2019-03-05)
Diffuse large B cell lymphoma (DLBCL) is a heterogeneous disease treated with anti-CD20-based immuno-chemotherapy (R-CHOP). We identified that low levels of GAPDH predict a poor response to R-CHOP treatment. Importantly, we demonstrated that GAPDHlow lymphomas use OxPhos metabolism and rely
Dries Vandamme et al.
Bioresource technology, 105, 114-119 (2011-12-21)
Microalgae hold great potential as a feedstock for biofuels or bulk protein or treatment of wastewater or flue gas. Realising these applications will require the development of a cost-efficient harvesting technology. Here, we explore the potential of flocculation induced by
Sandra Nilsson et al.
PloS one, 7(11), e46980-e46980 (2012-11-28)
During the intraerythrocytic development of Plasmodium falciparum, the malaria parasite remodels the host cell cytosol by inducing membranous structures termed Maurer's clefts and inserting parasite proteins into the red blood cell cytoskeleton and plasma membrane. Pf332 is the largest known
I Irwansyah et al.
Advanced materials (Deerfield Beach, Fla.), 27(4), 648-654 (2014-12-03)
Antimicrobial hydrogels are prepared based on the co-assembly of commercial Fmoc-phenylalanine and Fmoc-leucine, which act as the hydrogelator and antimicrobial building block, respectively. This co-assembled antimicrobial hydrogel is demonstrated to exhibit selective bactericidal activity for gram-positive bacteria while being biocompatible

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

  1. Is there an explanation between ACS grade vs. Reagent Plus for sodium carbonate part number 223530 compared to S2127?

    1 answer
    1. ACS stands for the American Chemical Society, which sets specifications for chemicals. Purchasing an ACS grade chemical means the product meets the minimum requirements set by the American Chemical Society, ensuring consistency across suppliers. Reagent Plus is our own grade with its own specifications, making product comparison essential when purchasing from suppliers with their own specifications. Purchasing products with ACS specifications simplifies the comparison process.

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service