Merck
  • Development and application of a non-targeted extraction method for the analysis of migrating compounds from plastic baby bottles by GC-MS.
Development and application of a non-targeted extraction method for the analysis of migrating compounds from plastic baby bottles by GC-MS.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2014-11-20)
Matthias Onghena, Els van Hoeck, Philippe Vervliet, Marie Louise Scippo, Coraline Simon, Joris van Loco, Adrian Covaci
ABSTRACT
In 2011, the European Union prohibited the production of polycarbonate (PC) baby bottles due to the toxic effects of the PC monomer bisphenol-A. Therefore, baby bottles made of alternative materials, e.g. polypropylene (PP) or polyethersulphone (PES), are currently marketed. The principal aim of the study was the identification of major compounds migrating from baby bottles using a liquid-liquid extraction followed by GC/MS analysis. A 50% EtOH in water solution was selected as a simulant for milk. After sterilisation of the bottle, three migration experiments were performed during 2 h at 70°C. A non-targeted liquid-liquid extraction with ethyl acetate-n-hexane (1:1) was performed on the simulant samples. Identification of migrants from 24 baby bottles was done using commercially available WILEY and NIST mass spectra libraries. Differences in the migrating compounds and their intensities were observed between the different types of plastics, but also between the same polymer from a different producer. Differences in the migration patterns were perceived as well between the sterilisation and the migrations and within the different migrations. Silicone, Tritan™ and PP exhibited a wide variety of migrating compounds, whereas PES and polyamide (PA) showed a lower amount of migrants, though sometimes in relatively large concentrations (azacyclotridecan-2-one up to 250 µg kg⁻¹). Alkanes (especially in PP bottles), phthalates (dibutylphthalate in one PP bottle (±40 µg kg⁻¹) and one silicone bottle (±25 µg kg⁻¹); diisobutylphthalate in one PP (±10 µg kg⁻¹), silicone (up to ±80 µg kg⁻¹); and Tritan™ bottle (±30 µg kg⁻¹)), antioxidants (Irgafos 168, degradation products of Irganox 1010 and Irganox 1076), etc. were detected for PP, silicone and Tritan™ bottles. Although the concentrations were relatively low, some compounds not authorised by European Union Regulation No. 10/2011, such as 2,4-di-tert-butylphenol (10-100 µg kg⁻¹) or 2-butoxyethyl acetate (about 300 µg kg⁻¹) were detected. Migrating chemicals were identified as confirmed (using a standard) or as tentative (further confirmation required).
Product Description
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Melting point standard 47-49°C, analytical standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
4-tert-Octylphenol, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
2,6-Diisopropylnaphthalene, AldrichCPR
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Eucalyptol, tested according to Ph. Eur.
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
2-Cyclohexen-1-one, ≥95%
Supelco
Myristic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Diisobutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Myristic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Supelco
o-Cresol, analytical standard
Supelco
Palmitic acid, analytical standard
Supelco
Acetic acid, analytical standard
Supelco
p-Cresol, analytical standard
Supelco
Methanol, analytical standard
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Supelco
2-Ethylhexyl acrylate, analytical standard
Supelco
Myristic acid, analytical standard
Supelco
Eucalyptol, analytical standard
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Supelco
tert-Butyl methyl ether, analytical standard