Skip to Content
Merck
All Photos(1)

Key Documents

442858

Supelco

4-tert-Octylphenol

analytical standard

Synonym(s):

4-(1,1,3,3-Tetramethylbutyl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2C(CH3)2C6H4OH
CAS Number:
Molecular Weight:
206.32
Beilstein:
513992
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

175 °C/30 mmHg (lit.)

mp

79-82 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

CC(C)(C)CC(C)(C)c1ccc(O)cc1

InChI

1S/C14H22O/c1-13(2,3)10-14(4,5)11-6-8-12(15)9-7-11/h6-9,15H,10H2,1-5H3

InChI key

ISAVYTVYFVQUDY-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

Application

4-tert-Octylphenol has been used as a reference standard for the determination of the analyte in water samples using ultra-high-performance liquid chromatography/tandem mass spectrometry (UHPLC/MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

296.6 °F

Flash Point(C)

147 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct analysis of trace level bisphenol A, octylphenols and nonylphenol in bottled water and leached from bottles by ultra?high?performance liquid chromatography/tandem mass spectrometry.
Wang J and Schnute CW
Rapid Communications in Mass Spectrometry, 24(17), 2605-2610 (2010)
Mei-Hui Li
Environmental toxicology and chemistry, 31(4), 843-850 (2012-01-27)
Surfactants are a major class of emerging pollutants widely used in large quantities in everyday life and commonly found in surface waters worldwide. Freshwater planarian was selected to examine the effects of different surfactants by measuring mortality, mobility, and membrane-bound
Tadashi Toyama et al.
Environmental science & technology, 45(15), 6524-6530 (2011-07-09)
We investigated biodegradation of technical nonylphenol (tNP) in Phragmites australis rhizosphere sediment by conducting degradation experiments using sediments spiked with tNP. Accelerated tNP removal was observed in P. australis rhizosphere sediment, whereas tNP persisted in unvegetated sediment without plants and
Qingxiang Zhou et al.
Talanta, 85(3), 1598-1602 (2011-08-03)
Present study described a simple, sensitive, and viable method for the determination of bisphenol A, 4-n-nonylphenol and 4-tert-octylphenol in water samples using temperature-controlled ionic liquid dispersive liquid-phase microextraction coupled to high performance liquid chromatography-fluorescence detector. In this experiment, 1-octyl-3-methylimidazolium hexafluorophosphate
Wen-Ling Chen et al.
Talanta, 89, 237-245 (2012-01-31)
This study developed and validated a method of measuring the feminizing chemicals 4-tert-octylphenol, 4-nonylphenol, nonylphenol monoethoxycarboxylate (NP(1)EC), nonylphenol monoethoxylate (NP(1)EO), nonylphenol diethoxylate (NP(2)EO), estrone, 17β-estradiol, estriol, 17α-ethinyl estradiol and bisphenol A in river water, sediment, and tissue using ultra-high performance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service